Merck
  • A super-ecliptic, pHluorin-mKate2, tandem fluorescent protein-tagged human LC3 for the monitoring of mammalian autophagy.
A super-ecliptic, pHluorin-mKate2, tandem fluorescent protein-tagged human LC3 for the monitoring of mammalian autophagy.
PloS one (2014-10-24)
Isei Tanida, Takashi Ueno, Yasuo Uchiyama
ABSTRACT
Tandem fluorescent protein-tagged LC3s that were comprised of a protein tag that emits green fluorescence (e.g., EGFP or mWasabi) fused with another tag that emits red fluorescence (e.g. mCherry or TagRFP) were used for monitoring the maturation step of mammalian autophagosomes. A critical point for this tandem fluorescent-tagged LC3 was the sensitivity of green fluorescence at an acidic pH. EGFP and mWasabi continue to emit a weak, but significant, fluorescence at a pH of approximately 6. To overcome this issue, we focused on super-ecliptic pHluorin, which is a more pH-sensitive GFP variation. The green fluorescence of EGFP and mWasabi in the cells was still observed at weakly acidic levels (pH 6.0-6.5). In contrast, the fluorescence of pHluorin was more significantly quenched at pH 6.5, and was almost completely abolished at pH 5.5-6.0, indicating that pHluorin is more suitable for use in a tandem fluorescent protein-tag for monitoring autophagy. A pHluorin-mKate2 tandem fluorescence protein showed pH-sensitive green fluorescence and pH-resistant far-red fluorescence. We therefore generated expression plasmids for pHluorin-mKate2-tagged human LC3 (PK-hLC3), which could be used as a modifier for LC3-lipidation. The green and far-red fluorescent puncta of PK-hLC3 were increased under starvation conditions. Puncta that were green-negative, but far-red positive, were increased when autolysosomes accumulated, but few puncta of the mutant PK-hLC3ΔG that lacked the carboxyl terminal Gly essential for autophagy were observed in the cells under the same conditions. These results indicated that the PK-hLC3 were more appropriate for the pH-sensitive monitoring of the maturation step of autophagosomes.
Sigma-Aldrich
Chloroquine diphosphate salt, powder or crystals, 98.5-101.0% (EP), meets EP testing specifications
Supelco
Ammonium chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Ammonium chloride, 99.99% trace metals basis
Sigma-Aldrich
Ammonium chloride, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Ammonium chloride, 99.998% trace metals basis
Sigma-Aldrich
Ammonium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Ammonium chloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
Ammonium chloride, Molecular Biology, suitable for cell culture, ≥99.5%
Sigma-Aldrich
Rapamycin, Ready Made Solution, 2.5 mg/mL in DMSO (2.74 mM), from Streptomyces hygroscopicus
Sigma-Aldrich
Ammonium-14N chloride, 99.99 atom % 14N, 15N-depleted, 99% (CP)
Supelco
Ammonium ion solution for ISE, 1000 mg/kg N, analytical standard (for ion-selective electrodes)
Supelco
Rapamycin, VETRANAL®, analytical standard
Supelco
Chloroquine phosphate, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
ANTI-FLAG® M2 antibody, Mouse monoclonal, 1 mg/mL, clone M2, affinity isolated antibody, buffered aqueous solution (50% glycerol, 10 mM sodium phosphate, and 150 mM NaCl, pH 7.4)