Merck
  • Simultaneous targeting of two ligand-binding sites on VEGFR2 using biparatopic Affibody molecules results in dramatically improved affinity.
Simultaneous targeting of two ligand-binding sites on VEGFR2 using biparatopic Affibody molecules results in dramatically improved affinity.
Scientific reports (2014-12-18)
Filippa Fleetwood, Susanne Klint, Martin Hanze, Elin Gunneriusson, Fredrik Y Frejd, Stefan Ståhl, John Löfblom
ABSTRACT
Angiogenesis plays an important role in cancer and ophthalmic disorders such as age-related macular degeneration and diabetic retinopathy. The vascular endothelial growth factor (VEGF) family and corresponding receptors are regulators of angiogenesis and have been much investigated as therapeutic targets. The aim of this work was to generate antagonistic VEGFR2-specific affinity proteins having adjustable pharmacokinetic properties allowing for either therapy or molecular imaging. Two antagonistic Affibody molecules that were cross-reactive for human and murine VEGFR2 were selected by phage and bacterial display. Surprisingly, although both binders independently blocked VEGF-A binding, competition assays revealed interaction with non-overlapping epitopes on the receptor. Biparatopic molecules, comprising the two Affibody domains, were hence engineered to potentially increase affinity even further through avidity. Moreover, an albumin-binding domain was included for half-life extension in future in vivo experiments. The best-performing of the biparatopic constructs demonstrated up to 180-fold slower dissociation than the monomers. The new Affibody constructs were also able to specifically target VEGFR2 on human cells, while simultaneously binding to albumin, as well as inhibit VEGF-induced signaling. In summary, we have generated small antagonistic biparatopic Affibody molecules with high affinity for VEGFR2, which have potential for both future therapeutic and diagnostic purposes in angiogenesis-related diseases.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Pricing and availability is not currently available.
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Pricing and availability is not currently available.
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Pricing and availability is not currently available.
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, ~6 M in H2O, for amino acid analysis
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Pricing and availability is not currently available.
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Pricing and availability is not currently available.
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, ≥98% (HPLC), powder
Pricing and availability is not currently available.
Sigma-Aldrich
L-Thyroxine sodium salt pentahydrate, γ-irradiated, powder, BioXtra, suitable for cell culture
Pricing and availability is not currently available.
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Pricing and availability is not currently available.
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Pricing and availability is not currently available.
Valine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Alanine, European Pharmacopoeia (EP) Reference Standard
Pricing and availability is not currently available.
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Pricing and availability is not currently available.
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Pricing and availability is not currently available.
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Pricing and availability is not currently available.
USP
Levothyroxine, United States Pharmacopeia (USP) Reference Standard
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Valine, ≥97%
Pricing and availability is not currently available.
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Pricing and availability is not currently available.