Merck
  • Protective effects of epigallocatechin gallate (EGCG) on streptozotocin-induced diabetic nephropathy in mice.
Protective effects of epigallocatechin gallate (EGCG) on streptozotocin-induced diabetic nephropathy in mice.
Acta histochemica (2014-08-27)
Sang Pil Yoon, Young Hee Maeng, Ran Hong, Byung Rai Lee, Chong Gue Kim, Hyun Lee Kim, Jong Hoon Chung, Byung Chul Shin
ABSTRACT
There is increasing evidence suggesting that antioxidants in green tea extracts may protect kidneys on the progression of end-stage renal disease. We investigated the protective impacts of (-)-epigallocatechin 3-O-gallate (EGCG) against streptozotocin (STZ)-induced diabetic nephropathy in mice. The mice were divided into 5 groups (n=10 per group): control (saline, i.p.), STZ (200mg/kg, i.p.), EGCG50 (50mg/kg, S.Q.), EGCG100 (100mg/kg, S.Q.), and EGCG200 (200mg/kg, S.Q.). Animals were sacrificed at scheduled times after EGCG administration and then quantitative and qualitative analysis were performed. Compared with the control group, the STZ group showed an increase in levels of blood glucose, blood urea nitrogen, creatinine and urine protein amounts with a decrease in body weight. All the above parameters were significantly reversed with EGCG treatment, especially in the EGCG100 group. After STZ injection, there was a mesangial proliferation with increased renal osteopontin accumulation and its protein expression in the glomeruli and the proximal tubules. Mice kidneys after EGCG-treatment showed a reduced expression of above parameters and relatively improved histopathological findings. These results indicated that EGCG 100mg/kg might provide an effective protection against STZ-induced diabetic nephropathy in mice by osteopontin suppression.
Product Description
Supelco
(−)-Epigallocatechin gallate, analytical standard
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥80% (HPLC), from green tea
Sigma-Aldrich
(−)-Epigallocatechin gallate, ≥95%
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Supelco
Epigallocatechin gallate, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Creatinine, Pharmaceutical Secondary Standard; Certified Reference Material
Epigallocatechin gallate, primary reference standard