Merck
  • The role of fillers and sodium metabisulfite on drug release from aged polyox tablets.
The role of fillers and sodium metabisulfite on drug release from aged polyox tablets.
Drug development and industrial pharmacy (2013-08-22)
Saeed Shojaee, Ali Nokhodchi, Iain Cumming
ABSTRACT
Polyethylene oxides (PEOs) are extensively used to control the release rate of drugs from matrices. Unfortunately, polyox polymers are prone to oxidation under high temperature and relative humidity. The aim of this study was to investigate the effect of sodium metabisulfite as an antioxidant to overcome the drug release changes from polyox matrices (PEO 301 and 303) when stored at 40 °C. The effect of different types of fillers (lactose, mannitol and dicalcium phosphate dihydrate) on stability of diltiazem HCl release profiles was also investigated. Generally, the presence of sodium metabisulfite stabilized the release of drug from PEO matrices stored at 40 °C for 8 weeks. Whilst the absence of metabisulfite caused an increase in drug release from polyox matrices when stored at 40 °C. The results indicate that all three concentrations (0.25, 0.5 and 1% w/w) of sodium metabisulfite were able to overcome structural changes of polyox samples hence stabilizing the drug release. The results also showed that the incorporation of fillers in polyox matrices reduced the sensitivity of drug release when stored at elevated temperature. This indicates that when these excipients were used there was no need to incorporate additional antioxidant. DSC results showed that there was no difference in the melting points of fresh polyox samples and aged polyox samples containing sodium metabisulfite, whereas the melting point of aged polyox samples without sodium metabisulfite were lower than fresh polyox samples. This indicates that the presence of metabisulfite is essential to stabilize polyox samples.
Product Description
Mannitol, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Sodium metabisulfite, tested according to Ph. Eur.
Sigma-Aldrich
D-Lactose monohydrate, ACS reagent
Diltiazem hydrochloride, European Pharmacopoeia (EP) Reference Standard
USP
Sodium metabisulfite, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Sigma-Aldrich
D-Lactose monohydrate, ≥98.0% (HPLC)
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Lactose monohydrate, ≥99% (HPLC), BioUltra
USP
Diltiazem hydrochloride, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Sigma-Aldrich
D-Lactose monohydrate, tested according to Ph. Eur.
Sigma-Aldrich
Sodium metabisulfite, puriss., meets analytical specification of Ph. Eur., BP, NF, FCC, E223, dry, 97-100.5%
Supelco
Sodium metabisulfite, analytical standard
Sigma-Aldrich
Sodium metabisulfite, reagent grade, 97%
Supelco
Sodium Metabisulfite, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Sodium metabisulfite, ReagentPlus®, ≥99%
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
D-Mannitol, ACS reagent
USP
Lactose monohydrate, United States Pharmacopeia (USP) Reference Standard
Supelco
Lactose (Monohydrate), Pharmaceutical Secondary Standard; Certified Reference Material
Diltiazem for system suitability, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
(+)-cis-Diltiazem hydrochloride, ≥99% (HPLC)