Merck
  • Nanostructure synthesis at the solid-water interface: spontaneous assembly and chemical transformations of tellurium nanorods.
Nanostructure synthesis at the solid-water interface: spontaneous assembly and chemical transformations of tellurium nanorods.
Chemphyschem : a European journal of chemical physics and physical chemistry (2014-08-01)
T P Vinod, Natalya Froumin, Dimitry Mogiliansky, Leila Zeiri, Vladimir Ezersky, Raz Jelinek
ABSTRACT
Bottom-up synthesis offers novel routes to obtain nanostructures for nanotechnology applications. Most self-assembly processes are carried out in three dimensions (i.e. solutions); however, the large majority of nanostructure-based devices function in two dimensions (i.e. on surfaces). Accordingly, an essential and often cumbersome step in bottom-up applications involves harvesting and transferring the synthesized nanostructures from the solution onto target surfaces. We demonstrate a simple strategy for the synthesis and chemical transformation of tellurium nanorods, which is carried out directly at the solid-solution interface. The technique involves binding the nanorod precursors onto amine-functionalized surfaces, followed by in situ crystallization/oxidation. We show that the surface-anchored tellurium nanorods can be further transformed in situ into Ag2Te, Cu2Te, and SERS-active Au-Te nanorods. This new approach offers a way to construct functional nanostructures directly on surfaces.
Sigma-Aldrich
Sodium borohydride, powder, ≥98.0%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98.0%
Sigma-Aldrich
Sodium borohydride, ReagentPlus®, 99%
Sigma-Aldrich
Sodium borohydride, purum p.a., ≥96% (gas-volumetric)
Sigma-Aldrich
Sodium borohydride, granular, 10-40 mesh, 98%
Sigma-Aldrich
Sodium borohydride, caplets (18 × 10 × 8 mm), 98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, ≥98%
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, 99%
Sigma-Aldrich
Sodium borohydride, granular, 99.99% trace metals basis
Sigma-Aldrich
Hydrazine monohydrate, N2H4 64-65 %, reagent grade, ≥97%
Sigma-Aldrich
Hydrazine hydrate solution, puriss. p.a., 24-26% in H2O (RT)
Sigma-Aldrich
(3-Aminopropyl)triethoxysilane, packaged for use in deposition systems, ≥98%
Sigma-Aldrich
Rhodamine 6G, Dye content ~95 %
Sigma-Aldrich
Rhodamine 6G, Dye content 99 %
Sigma-Aldrich
Sodium borohydride solution, ~12 wt. % in 14 M NaOH
Sigma-Aldrich
Sodium borohydride solution, 2.0 M in triethylene glycol dimethyl ether
Sigma-Aldrich
Hydrazine hydrate, reagent grade, N2H4 50-60 %
Supelco
Rhodamine 6G, analytical standard
Sigma-Aldrich
Rhodamine 6G, suitable for fluorescence, BioReagent
Sigma-Aldrich
Tellurium, granular, −5-+50 mesh, 99.99% trace metals basis
Sigma-Aldrich
Tellurium, shot, ≤2 mm, 99.999% trace metals basis
Sigma-Aldrich
Tellurium, powder, −30 mesh, 99.997% trace metals basis
Sigma-Aldrich
Tellurium, powder, −200 mesh, 99.8% trace metals basis
Sigma-Aldrich
Tellurium, 99.999% trace metals basis
Sigma-Aldrich
VenPure® SF, powder