Merck
  • Chloride channel ClC-5 binds to aspartyl aminopeptidase to regulate renal albumin endocytosis.
Chloride channel ClC-5 binds to aspartyl aminopeptidase to regulate renal albumin endocytosis.
American journal of physiology. Renal physiology (2015-01-15)
Aven Lee, Craig Slattery, David J Nikolic-Paterson, Deanne H Hryciw, Sherwin Wilk, Elizabeth Wilk, Yuan Zhang, Valentina A Valova, Phillip J Robinson, Darren J Kelly, Philip Poronnik
ABSTRACT
ClC-5 is a chloride/proton exchanger that plays an obligate role in albumin uptake by the renal proximal tubule. ClC-5 forms an endocytic complex with the albumin receptor megalin/cubilin. We have identified a novel ClC-5 binding partner, cytosolic aspartyl aminopeptidase (DNPEP; EC 3.4.11.21), that catalyzes the release of N-terminal aspartate/glutamate residues. The physiological role of DNPEP remains largely unresolved. Mass spectrometric analysis of proteins binding to the glutathione-S-transferase (GST)-ClC-5 C terminus identified DNPEP as an interacting partner. Coimmunoprecipitation confirmed that DNPEP and ClC-5 also associated in cells. Further experiments using purified GST-ClC-5 and His-DNPEP proteins demonstrated that the two proteins bound directly to each other. In opossum kidney (OK) cells, confocal immunofluorescence studies revealed that DNPEP colocalized with albumin-containing endocytic vesicles. Overexpression of wild-type DNPEP increased cell-surface levels of ClC-5 and albumin uptake. Analysis of DNPEP-immunoprecipitated products from rat kidney lysate identified β-actin and tubulin, suggesting a role for DNPEP in cytoskeletal maintenance. A DNase I inhibition assay showed a significant decrease in the amount of G actin when DNPEP was overexpressed in OK cells, suggesting a role for DNPEP in stabilizing the cytoskeleton. DNPEP was not present in the urine of healthy rats; however, it was readily detected in the urine in rat models of mild and heavy proteinuria (diabetic nephropathy and anti-glomerular basement membrane disease, respectively). Urinary levels of DNPEP were found to correlate with the severity of proteinuria. Therefore, we have identified another key molecular component of the albumin endocytic machinery in the renal proximal tubule and describe a new role for DNPEP in stabilizing the actin cytoskeleton.
Product Description
Supelco
4-tert-Octylphenol monoethoxylate solution, 10 μg/mL in acetone, analytical standard
Sigma-Aldrich
N-Hydroxynaphthalimide triflate, electronic grade, ≥99%
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
Hydrogen peroxide solution, ≥30%, for trace analysis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Millipore
Hydrogen peroxide solution, 3%, suitable for microbiology
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Supelco
Sodium chloride, reference material for titrimetry, certified by BAM, >99.5%
Sigma-Aldrich
Sodium chloride, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Hydrogen peroxide solution, tested according to Ph. Eur.
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride solution, 5 M
SAFC
Sodium chloride solution, 5 M
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Supelco
Hydrogen peroxide solution, 30 % (w/w), for ultratrace analysis
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)