Merck
  • Separation and structural elucidation of a new tadalafil analogue diethylaminopretadalafil included as an adulterant in a dietary supplement.
Separation and structural elucidation of a new tadalafil analogue diethylaminopretadalafil included as an adulterant in a dietary supplement.
Journal of pharmaceutical and biomedical analysis (2014-03-19)
Gaofei Zhang, Yue Yu, Xiaoou Wu, Jun Li
ABSTRACT
A new tadalafil analogue was detected and isolated from a dietary supplement, the structure was elucidated by means of nuclear magnetic resonance (NMR) and mass spectroscopy. The compound was determined to be diethylaminopretadalafil, which might be derived from a precursor in the synthesis of tadalafil.
Product Description
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Supelco
Acetonitrile, analytical standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
USP
Residual Solvent Class 2 - Acetonitrile, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%
Sigma-Aldrich
Acetonitrile, HPLC Plus, ≥99.9%