Merck
  • Retrospective monitoring of long-term recreational and dependent cocaine use in toenail clippings/scrapings as an alternative to hair.
Retrospective monitoring of long-term recreational and dependent cocaine use in toenail clippings/scrapings as an alternative to hair.
Bioanalysis (2014-12-23)
Milena M Madry, Andrea E Steuer, Matthias Vonmoos, Boris B Quednow, Markus R Baumgartner, Thomas Kraemer
ABSTRACT
Toenails were assessed as an alternative matrix to hair for retrospective monitoring of cocaine consumption of recreational and dependent users. Results/methodology: Toenail clippings, scrapings and hair samples from recreational and dependent cocaine users were analyzed for cocaine and metabolites. Dependent users displayed significantly higher concentrations in hair and toenail samples compared to recreational users. Cocaine abstinence could be monitored in hair and toenail samples. One postmortem fingernail was analyzed in layers to investigate the cocaine and metabolite concentration profile. Highest concentrations were observed in the dorsal layer, being indicative of contamination. Having led to comparable results, toenails may be an alternative for retrospective monitoring of cocaine consumption/abstinence. Hair should remain the first choice for assessment of temporal evidence of drug intake.
Product Description
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Acetone, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Acetone, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Sigma-Aldrich
Acetone, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Acetone, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Acetone, histological grade, ≥99.5%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Acetone, natural, ≥97%
Sigma-Aldrich
Acetone, ≥99%, FCC, FG
Supelco
Acetone, analytical standard
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Acetone, ACS reagent, ≥99.5%
Supelco
Methanol, analytical standard
Sigma-Aldrich
β-D-Allose, rare aldohexose sugar
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard