Merck
  • Preparation and characterization of amphiphilic calixarene nanoparticles as delivery carriers for paclitaxel.
Preparation and characterization of amphiphilic calixarene nanoparticles as delivery carriers for paclitaxel.
Chemical & pharmaceutical bulletin (2015-03-12)
Zi-Ming Zhao, Yu Wang, Jin Han, Hui-Dong Zhu, Lin An
ABSTRACT
Two types of amphoteric calix[n]arene carboxylic acid (CnCA) derivative, i.e., calix[6]arene hexa-carboxylic acid (C6HCA) and calix[8]arene octo-carboxylic acid (C8OCA), were synthesized by introducing acetoxyls into the hydroxyls of calix[n]arene (n=6, 8). C6HCA and C8OCA nanoparticles (NPs) were prepared successfully using the dialysis method. CnCA NPs had regular spherical shapes with an average diameter of 180-220 nm and possessed negative charges of greater than -30 mV. C6HCA and C8OCA NPs were stable in 4.5% bovine serum albumin solutions and buffers (pH 5-9), with a low critical aggregation concentration value of 5.7 mg·L(-1) and 4.0 mg·L(-1), respectively. C6HCA and C8OCA NPs exhibited good paclitaxel (PTX) loading capacity, with drug loading contents of 7.5% and 8.3%, respectively. The overall in vitro release behavior of PTX from the CnCA NPs was sustained, and C8OCA NPs had a slower release rate compared with C6HCA NPs. These favorable properties of CnCA NPs make them promising nanocarriers for tumor-targeted drug delivery.
Sigma-Aldrich
Tetrahydrofuran, anhydrous, ≥99.9%, inhibitor-free
USP
Dehydrated Alcohol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Tetrahydrofuran, ACS reagent, ≥99.0%, contains 200-400 ppm BHT as inhibitor
Sigma-Aldrich
Tetrahydrofuran, contains 200-400 ppm BHT as inhibitor, ACS reagent, ≥99.0%
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Supelco
Acetonitrile(Neat), Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Dehydrated Alcohol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Tetrahydrofuran, inhibitor-free, suitable for HPLC, ≥99.9%
Supelco
Tetrahydrofuran, Selectophore, ≥99.5%
Supelco
Tetrahydrofuran, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
Tetrahydrofuran, analytical standard
Supelco
Residual Solvent - Acetonitrile(solution in DMSO), Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
Acetonitrile, suitable for HPLC, gradient grade, ≥99.9%