Merck
  • Toll-like receptor 2 and 4 expression in the pregnant and non-pregnant human uterine cervix.
Toll-like receptor 2 and 4 expression in the pregnant and non-pregnant human uterine cervix.
Journal of reproductive immunology (2014-12-04)
Behnia S Lashkari, Shahida Shahana, Dilly O Anumba
ABSTRACT
Pelvic infections and sexually transmitted diseases place a burden on health resources and may be associated with premature birth. The mechanisms by which the female reproductive tract (FRT) combats these infections remain ill understood, but are likely to involve the pattern recognition Toll-like receptors (TLR). We sought to compare the expression of TLR-2 and -4 by human pregnant and non-pregnant ectocervical epithelium as a prelude to the investigation of the function of these receptors in this tissue during pregnancy. Using the techniques of reverse-transcriptase polymer chain reaction (RT-PCR) and immunohistochemistry, the gene and protein expression of TLR-2 and -4 were studied in the biopsies of ectocervix obtained from non-pregnant premenopausal women (n=21) undergoing hysterectomy, women in the first trimester of pregnancy undergoing non-medically indicated suction pregnancy termination (n=6), and women at term undergoing elective caesarean section (n=11). The expression of TLR2 and TLR4 genes and proteins were upregulated in early and late pregnant ectocervical epithelium, compared with non-pregnant tissue. These findings suggest that the upregulation of TLR2 and TLR4 in the lower FRT may play a key role in the modulation of the innate immune and inflammatory mechanisms of the ectocervix during pregnancy, interacting with other neuroendocrine factors.
Sigma-Aldrich
2-Methylbutane, anhydrous, ≥99%
Supelco
2-Methylbutane, analytical standard
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, ReagentPlus®, ≥99%
Sigma-Aldrich
2-Methylbutane, suitable for HPLC, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, ≥99.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis