Merck
  • Creatine reduces hepatic TG accumulation in hepatocytes by stimulating fatty acid oxidation.
Creatine reduces hepatic TG accumulation in hepatocytes by stimulating fatty acid oxidation.
Biochimica et biophysica acta (2014-09-11)
Robin P da Silva, Karen B Kelly, Kelly-Ann Leonard, René L Jacobs
ABSTRACT
Non-alcoholic fatty liver disease encompasses a wide spectrum of liver damage including steatosis, non-alcoholic steatohepatitis, fibrosis and cirrhosis. We have previously reported that creatine supplementation prevents hepatic steatosis and lipid peroxidation in rats fed a high-fat diet. In this study, we employed oleate-treated McArdle RH-7777 rat hepatoma cells to investigate the role of creatine in regulating hepatic lipid metabolism. Creatine, but not structural analogs, reduced cellular TG accumulation in a dose-dependent manner. Incubating cells with the pan-lipase inhibitor diethyl p-nitrophenylphosphate (E600) did not diminish the effect of creatine, demonstrating that the TG reduction brought about by creatine does not depend on lipolysis. Radiolabeled tracer experiments indicate that creatine increases fatty acid oxidation and TG secretion. In line with increased fatty acid oxidation, mRNA analysis revealed that creatine-treated cells had increased expression of PPARα and several of its transcriptional targets. Taken together, this study provides direct evidence that creatine reduces lipid accumulation in hepatocytes by the stimulation of fatty acid oxidation and TG secretion.
Sigma-Aldrich
Oleic acid, meets analytical specification of Ph, Eur., 65.0-88.0% (GC)
Sigma-Aldrich
Oleic acid, natural, FCC
Sigma-Aldrich
2-Propanol, Laboratory Reagent, ≥99.5%
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
Isopropyl alcohol, meets USP testing specifications
Supelco
2-Propanol, analytical standard
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.9%
Sigma-Aldrich
2-Propanol, puriss. p.a., ACS reagent, ≥99.8% (GC)
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%
Sigma-Aldrich
2-Propanol, ACS reagent, ≥99.5%
USP
2-Propanol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Oleic acid, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Supelco
Oleic acid, Selectophore, ≥99%
Supelco
Oleic acid, analytical standard
Sigma-Aldrich
Oleic acid, technical grade, 90%
Supelco
2-Propanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Propanol, suitable for HPLC, 99.5%
Sigma-Aldrich
Oleic acid, ≥99% (GC)
Oleic acid, European Pharmacopoeia (EP) Reference Standard
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG