Merck
  • Inhibition of calcium-calmodulin complex formation by vasorelaxant basic dipeptides demonstrated by in vitro and in silico analyses.
Inhibition of calcium-calmodulin complex formation by vasorelaxant basic dipeptides demonstrated by in vitro and in silico analyses.
Biochimica et biophysica acta (2014-08-06)
Thanutchaporn Kumrungsee, Tomomi Saiki, Sayaka Akiyama, Kentaro Nakashima, Mitsuru Tanaka, Yutaro Kobayashi, Toshiro Matsui
ABSTRACT
Tryptophan-histidine (Trp-His) was found to suppress the activity of the Ca²⁺/calmodulin (CaM)-dependent protein kinases II (CaMKII), which requires the Ca²⁺-CaM complex for an initial activation. In this study, we attempted to clarify whether Trp-His inhibits Ca²⁺-CaM complex formation, a CaMKII activator. The ability of Trp-His and other peptides to inhibit Ca²⁺-CaM complex formation was investigated by a Ca²⁺-encapsulation fluorescence assay. The peptide-CaM interactions were illustrated by molecular dynamic simulation. We showed that Trp-His inhibited Ca²⁺-CaM complex formation with a 1:1 binding stoichiometry of the peptide to CaM, considering that Trp-His reduced Hill coefficient of Ca²⁺-CaM binding from 2.81 to 1.92. His-Trp also showed inhibitory activity, whereas Trp+His, 3-methyl His-Trp, and Phe-His did not show significant inhibitory activity, suggesting that the inhibitory activity was due to a peptide skeleton (irrespective of the sequence), a basic amino acid, a His residue, the N hydrogen atom of its imidazole ring, and Trp residue. In silico studies suggested the possibility that Trp-His and His-Trp interacted with the Ca²⁺-binding site of CaM by forming hydrogen bonds with key Ca²⁺-binding residues of CaM, with a binding free energy of -49.1 and -68.0 kJ/mol, respectively. This is the first study demonstrating that the vasoactive dipeptide Trp-His possesses inhibitory activity against Ca²⁺-CaM complex formation, which may elucidate how Trp-His inhibited CaMKII in a previous study. The results provide a basic idea that could lead to the development of small peptides binding with high affinity to CaM and inhibiting Ca²⁺-CaM complex formation in the future.
Product Description
Supelco
L-Tryptophan, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Sigma-Aldrich
L-Tryptophan, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Tryptophan, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
L-Tryptophan, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 99.0-101.0%
Supelco
L-Histidine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
L-Histidine, Pharmaceutical Secondary Standard; Certified Reference Material
SAFC
L-Histidine
Sigma-Aldrich
Gly-Gly, BioXtra, ≥99.0% (NT)
Sigma-Aldrich
Gly-Gly, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
L-Histidine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Gly-Gly, BioPerformance Certified, suitable for cell culture, ≥99%
Sigma-Aldrich
Gly-Gly, ≥99% (titration)
Sigma-Aldrich
L-Histidine, suitable for cell culture, meets EP, USP testing specifications, from non-animal source
Sigma-Aldrich
L-Histidine, ReagentPlus®, ≥99% (TLC)