Merck
  • Targeting cancer cells via the reactive oxygen species-mediated unfolded protein response with a novel synthetic polyphenol conjugate.
Targeting cancer cells via the reactive oxygen species-mediated unfolded protein response with a novel synthetic polyphenol conjugate.
Clinical cancer research : an official journal of the American Association for Cancer Research (2014-06-19)
Soon Young Shin, Jong Min Lee, Mi So Lee, Dongsoo Koh, Hyeryoung Jung, Yoongho Lim, Young Han Lee
ABSTRACT
The selective killing of tumor cells is an important strategy for cancer therapeutics. The aim of this study was to develop a novel antitumor agent that is safe for normal cells with the ability to selectively target cancer cells. On the basis of quantitative structure-activity relationship, we synthesized a novel polyphenol conjugate (E)-3-(3,5-dimethoxyphenyl)-1-(2-methoxyphenyl)prop-2-en-1-one (DPP-23). We evaluated the effect of DPP-23 on proliferation, cell cycle, and apoptosis in various tumor cells. We also assessed molecular targets of DPP-23 using genome-wide expression profiling by DNA microarray and real-time PCR array systems. DPP-23 effectively inhibited the growth of cancer cells in vitro and in vivo (xenografts in Balb/c nude mice). At a molecular level, DPP-23 targeted the unfolded protein response (UPR) in the endoplasmic reticulum (ER) through the production of reactive oxygen species (ROS) in cancer cells, but not in normal cells, resulting in selective killing of tumor cells via caspase-dependent apoptosis. The selective generation of ROS in cancer cells could be an attractive strategy for the selective killing of cancer cells, while maintaining negligible cytotoxicity to normal cells. DPP-23 represents a promising novel therapeutic agent for the selective production of ROS in cancer cells.
Product Description
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
L-Glutathione reduced, suitable for cell culture, BioReagent, ≥98.0%, powder
Sigma-Aldrich
L-Glutathione reduced, ≥98.0%
Sigma-Aldrich
L-Glutathione reduced, BioXtra, ≥98.0%
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
MISSION® esiRNA, targeting human BECN1
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, ≥99.5% (GC)
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Becn1
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Mia1
Supelco
Glutathione, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
MISSION® esiRNA, targeting human ERN1