Merck
  • Dilong prevents the high-KCl cardioplegic solution administration-induced apoptosis in H9c2 cardiomyoblast cells mediated by MEK.
Dilong prevents the high-KCl cardioplegic solution administration-induced apoptosis in H9c2 cardiomyoblast cells mediated by MEK.
The American journal of Chinese medicine (2014-12-09)
Chien-Kuo Han, Wei-Wen Kuo, Chia-Yao Shen, Tung-Sheng Chen, Peiying Pai, Chang-Hai Tsai, Feng-Yueh Lo, Da-Tong Ju, Chih-Yang Huang
ABSTRACT
Infusion of high-KCl cardioplegic solution (High-KCS) is the most common method used to induce asystole before cardiac surgery. However, our previous study showed the High-KCS can cause the apoptosis of cardiomyocytes in patients who were administered High-KCS prior to undergoing coronary artery bypass graft (CABG) to treat coronary artery disease (CAD). Therefore, it is urgent today to find a complementary medicine to reduce this damage. Dilong (earthworm) has been used as a traditional medicine in China for several thousand years, and extract from the dilong has been empirically used in Asia for the treatment of vascular disorders. In this study, we applied dilong extract to reduce myocardial cell damage from High-KCS infusion and further investigated the mechanisms. H9c2 cardiomyoblast cells were cultured in serum-free medium for 4 h and then treated with dilong at 31.25, 62.5, 125, and 250 mg/mL for 24 h, which was then followed by High-KCS treatment for 3 h to detect the protective mechanisms of dilong behind cardiomyocyte apoptosis and cardiac fibrosis. Cells were harvested for MTT assay, TUNEL assay, and western blot analysis. We found that High-KCS-induced cardiomyocyte apoptosis, enhanced the protein level of pro-apoptotic Bad, released cytochrome c, and activated caspase-3 in H9c2 cells. The IGF-I/IGF-IR/ERK pathway involved in non-cardiomyocyte proliferation, and the expression/activation of uPA, Sp-1 and CTGF, which are implicated in the development of cardiac fibrosis were up-regulated, but the Akt for cardiomyocyte survival was greatly deactivated in postcardioplegic H9c2 cardiomyoblast cells. However, dilong was highly protective and totally reversed the apoptosis and cardiac fibrosis effects induced by High-KCS. Chemical inhibitors P38 (SB203580), JNK (SP600125), MEK (U0126), IGF-1 (AG1024), and PI3K (LY294002) were applied to investigate which is the mediator for dilong attenuated High-KCS stimulated caspase 3 activation. MEK (U0126) inhibitor completely blocked dilong inhibited caspase 3 activation in High-KCS treated H9c2 cells. The MEK siRNA was further applied to knockdown MEK to confirm our finding. We found dilong worked through MEK to inhibit caspase 3 activity induced by High-KCS in H9c2 cells. Furthermore, we used the pure component of dilong, Lumbrokinase, to block the High-KCS effect. Using the microscope to observe the cell viability, we found Lumbrokinase could reverse the High-KCS effect. Lumbrokinase could also reduce the protein levels of caspase 8, caspase 9, and caspase 3, and enhance the survival related proteins PI3K/Akt and Bcl2. These results demonstrate that dilong could be used as a potential agent to block the side effects caused by High-KCS in CABG surgery patients.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Mercaptoethanol, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Sigma-Aldrich
2-Mercaptoethanol, Molecular Biology, suitable for electrophoresis, suitable for cell culture, BioReagent, 99% (GC/titration)
Sigma-Aldrich
2-Mercaptoethanol, ≥99.0%
Supelco
2-Mercaptoethanol, derivatization grade (HPLC), LiChropur, ≥99.0% (GC)
USP
Glycerin, United States Pharmacopeia (USP) Reference Standard
Supelco
Glycerol, analytical standard
Supelco
Methyl Ethyl Ketone, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
2-Butanone, ACS reagent, ≥99.0%
Sigma-Aldrich
Glycerol, ReagentPlus®, ≥99.0% (GC)
Sigma-Aldrich
Glycerol, ACS reagent, ≥99.5%
Sigma-Aldrich
Glycerol, FCC, FG
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ≥98.0% (KT)
Sigma-Aldrich
Glycerol, ≥99.5%
Sigma-Aldrich
Glycerol, Molecular Biology, ≥99.0%
Sigma-Aldrich
Ethylenediaminetetraacetic acid solution, 0.02% in DPBS (0.5 mM), sterile-filtered, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerol, BioXtra, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Glycerin, meets USP testing specifications
Sigma-Aldrich
Glycerol, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for electrophoresis, ≥99% (GC)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid disodium salt solution, BioUltra, pH 8.0, ~0.5 M in H2O
Supelco
2-Butanone, analytical standard
Sigma-Aldrich
Glycerol, tested according to Ph. Eur., anhydrous