Merck
  • A mass spectrometry-based method for differentiation of positional isomers of monosubstituted pyrazine N-oxides using metal ion complexes.
A mass spectrometry-based method for differentiation of positional isomers of monosubstituted pyrazine N-oxides using metal ion complexes.
Journal of mass spectrometry : JMS (2015-01-21)
Matías Butler, Gabriela M Cabrera
ABSTRACT
A series of 11 pairs of substituted pyrazine N-oxides, differing in the substituent position, were examined using electrospray ionization mass spectrometry (ESI-MS) in order to use spectra to assess the differentiation of positional isomers. For each compound, mass spectra were recorded with three different metal cations, namely calcium (II), copper (II) and aluminum (III), with characterization of the observed peaks. Differentiation between regioisomeric N-oxides has been achieved by comparison of the identity and relative intensities of the peaks originating from the adduct ions formed with the metal ions. Principal component analysis (PCA) has been employed to assist in the interpretation of the results obtained with each metal ion, exploring possible trends according to the nature and position of the substituent in the pyrazine N-oxide.
Sigma-Aldrich
Methanol, suitable for HPLC, gradient grade, suitable as ACS-grade LC reagent, ≥99.9%
Supelco
Methanol, analytical standard
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, ACS spectrophotometric grade, ≥99.9%
Sigma-Aldrich
Methanol, BioReagent, ≥99.93%
Sigma-Aldrich
Methanol, suitable for HPLC, ≥99.9%
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Sigma-Aldrich
Methanol, HPLC Plus, ≥99.9%
Supelco
Methanol, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
USP
Methyl alcohol, United States Pharmacopeia (USP) Reference Standard