Merck
  • Cell-penetrating antimicrobial peptides - prospectives for targeting intracellular infections.
Cell-penetrating antimicrobial peptides - prospectives for targeting intracellular infections.
Pharmaceutical research (2015-03-18)
Jesper S Bahnsen, Henrik Franzyk, Edward J Sayers, Arwyn T Jones, Hanne M Nielsen
ABSTRACT
To investigate the suitability of three antimicrobial peptides (AMPs) as cell-penetrating antimicrobial peptides. Cellular uptake of three AMPs (PK-12-KKP, SA-3 and TPk) and a cell-penetrating peptide (penetratin), all 5(6)-carboxytetramethylrhodamine-labeled, were tested in HeLa WT cells and analyzed by flow cytometry and confocal microscopy. Furthermore, the effects of the peptides on eukaryotic cell viability as well as their antimicrobial effect were tested. In addition, the disrupting ability of the peptides in the presence of bilayer membranes of different composition were analyzed. AMP uptake relative to penetratin was ~13% (PK-12-KKP), ~66% (SA-3) and ~50% (TPk). All four peptides displayed a punctate uptake pattern in HeLa WT cells with co-localization to lysosomes and no indication that clathrin-mediated endocytosis was the predominant uptake mechanism. TPk showed the highest antibacterial activity. SA-3 exhibited selective disruption of liposomes mimicking Gram-positive and Gram-negative membranes. PK-12-KKP is an unlikely candidate for targeting intracellular bacteria, as the eukaryotic cell-penetrating ability is poor. SA-3, affected the cellular viability to an unacceptable degree. TPk showed acceptable uptake efficiency, high antimicrobial activity and relatively low toxicity, and it is the best potential lead peptide for further development.
Product Description
Supelco
Glycerin, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥99.0% (TLC)
Sigma-Aldrich
N-Chlorosuccinimide, 98%
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Cholesterol, tested according to Ph. Eur.
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Supelco
2,2,2-Trifluoroethanol, analytical standard, suitable for NMR (spectroscopy)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
5(6)-Carboxytetramethylrhodamine, BioReagent, suitable for fluorescence, ≥85% (sum of isomers, HPCE)
Supelco
N,N-Dimethylformamide, analytical standard
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Glycerol solution, 83.5-89.5% (T)
Supelco
Sodium dodecyl sulfate, suitable for ion pair chromatography, LiChropur, ≥99.0%
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
N,N-Dimethylformamide, Molecular Biology, ≥99%
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ReagentPlus®, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Sigma-Aldrich
L-α-Phosphatidylethanolamine, dioleoyl, ≥99% (GC), ≥98% (TLC), lyophilized powder
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration), suitable for cell culture
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
2-Oleoyl-1-palmitoyl-sn-glycero-3-phosphocholine, ≥95.5% (GC), ≥98% (TLC)