Merck
Effect of curcumin in mice model of vincristine-induced neuropathy.
Pharmaceutical biology (2014-11-29)
Anand Babu, K G Prasanth, Bhaskar Balaji
ABSTRACT
Curcumin exhibits a wide spectrum of biological activities which include neuroprotective, antinociceptive, anti-inflammatory, and antioxidant activity. The present study evaluates the effect of curcumin in vincristine-induced neuropathy in a mice model. Vincristine sulfate (0.1 mg/kg, i.p. for 10 consecutive days) was administered to mice to induce neuropathy. Pain behavior was assessed at different days, i.e., 0, 7, 10, and 14 d. Sciatic nerve total calcium, superoxide dismutase (SOD), catalase (CAT), glutathione peroxidase (GPx), reduced glutathione (GSH), nitric oxide (NO), and lipid peroxidation (LPO) were also estimated after the 14th day of study. Pregabalin (10 mg/kg, p.o.) and curcumin (15, 30, and 60 mg/kg, p.o.) were administered for 14 consecutive days. Curcumin at 60 mg/kg significantly attenuated the vincristine-induced neuropathic pain manifestations in terms of thermal hyperalgesia (p < 0.001) and allodynia (p < 0.001); mechanical hyperalgesia (p < 0.001); functional loss (p < 0.001); and in the delayed phase of formalin test (p < 0.001). Curcumin at 30 and 60 mg/kg exhibited significant changes (p < 0.001) in antioxidant levels and in total calcium levels in vincristine-injected mice. Curcumin at 30 and 60 mg/kg dose levels significantly attenuated vincristine-induced neuropathy which may be due to its multiple actions including antinociceptive, calcium inhibitory, and antioxidant effect.
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Supelco
Phenol, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Phenol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Phosphoric acid-16O4 solution, 70 wt. % in D2O, 99.9 atom % 16O
Supelco
Phenol solution, 5000 μg/mL in methanol, certified reference material
Supelco
Phenol solution, certified reference material, 500 μg/mL in methanol
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Vincristine sulfate salt, 95.0-105.0% (HPLC), powder or crystals
Supelco
Phenol solution, 100 μg/mL in acetonitrile, PESTANAL®, analytical standard
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, BioUltra, ≥85% (T)
Sigma-Aldrich
Phosphoric acid, ≥85 wt. % in H2O, ≥99.999% trace metals basis
Sigma-Aldrich
Phosphoric acid, BioReagent, suitable for insect cell culture, 85%
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phosphoric acid, ACS reagent, ≥85 wt. % in H2O
Sigma-Aldrich
Phosphoric acid, 85 wt. % in H2O, 99.99% trace metals basis
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Supelco
Phenol, PESTANAL®, analytical standard
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
Phosphoric acid, crystalline, ≥99.999% trace metals basis
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, natural, 97%, FG
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)