Merck
  • Interactions between bupropion and 3,4-methylenedioxymethamphetamine in healthy subjects.
Interactions between bupropion and 3,4-methylenedioxymethamphetamine in healthy subjects.
The Journal of pharmacology and experimental therapeutics (2015-02-07)
Yasmin Schmid, Anna Rickli, Antonia Schaffner, Urs Duthaler, Eric Grouzmann, Cédric M Hysek, Matthias E Liechti
ABSTRACT
3,4-Methylenedioxymethamphetamine (MDMA; "ecstasy") is a popular recreational drug. The aim of the present study was to explore the role of dopamine in the psychotropic effects of MDMA using bupropion to inhibit the dopamine and norepinephrine transporters through which MDMA releases dopamine and norepinephrine by investigating. The pharmacodynamic and pharmacokinetic interactions between bupropion and MDMA in 16 healthy subjects were investigated using a double-blind, placebo-controlled, crossover design. Bupropion reduced the MDMA-induced elevations in plasma norepinephrine concentrations and the heart rate response to MDMA. In contrast, bupropion increased plasma MDMA concentrations and prolonged its subjective effects. Conversely, MDMA increased plasma bupropion concentrations. These results indicate a role for the transporter-mediated release of norepinephrine in the cardiostimulant effects of MDMA but do not support a modulatory role for dopamine in the mood effects of MDMA. These results also indicate that the use of MDMA during therapy with bupropion may result in higher plasma concentrations of both MDMA and bupropion and enhanced mood effects but also result in lower cardiac stimulation.
Sigma-Aldrich
D-Mannitol, tested according to Ph. Eur.
Millipore
D-Mannitol, ACS reagent, suitable for microbiology, ≥99.0%
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Supelco
Hydrogen chloride – 2-propanol solution, ~1.25 M HCl (T), derivatization grade (GC derivatization), LiChropur
Supelco
Hydrogen chloride – ethanol, ~1.25 M HCl, derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Supelco
Mannitol, Pharmaceutical Secondary Standard; Certified Reference Material
Supelco
D-Mannitol, ≥99.9999% (metals basis), for boron determination
Supelco
Hydrogen chloride – methanol solution, ~1.25 m HCl (T), derivatization grade (GC derivatization), LiChropur
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
D-Mannitol, ACS reagent
Mannitol, European Pharmacopoeia (EP) Reference Standard
USP
Mannitol, United States Pharmacopeia (USP) Reference Standard
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%