Merck
  • Pleiotropic effects of spongean alkaloids on mechanisms of cell death, cell cycle progression and DNA damage response (DDR) of acute myeloid leukemia (AML) cells.
Pleiotropic effects of spongean alkaloids on mechanisms of cell death, cell cycle progression and DNA damage response (DDR) of acute myeloid leukemia (AML) cells.
Cancer letters (2015-02-24)
Fabian Stuhldreier, Stefanie Kassel, Lena Schumacher, Sebastian Wesselborg, Peter Proksch, Gerhard Fritz
ABSTRACT
We investigated cytotoxic mechanisms evoked by the spongean alkaloids aaptamine (Aa) and aeroplysinin-1 (Ap), applied alone and in combination with daunorubicin, employing acute myeloid leukemia (AML) cells. Aa and Ap reduced the viability of AML cells in a dose dependent manner with IC50 of 10-20 µM. Ap triggered apoptotic cell death more efficiently than Aa. Both alkaloids increased the protein level of S139-phosphorylated H2AX (γH2AX), which however was independent of the induction of DNA damage. Expression of the senescence markers p21 and p16 was increased, while the phosphorylation level of p-Chk-2 was reduced following Aa treatment. As a function of dose, Aa and Ap protected or sensitized AML cells against daunorubicin. Protection by Aa was paralleled by reduced formation of ROS and lower level of DNA damage. Both Aa and Ap attenuated daunorubicin-stimulated activation of the DNA damage response (DDR) as reflected on the levels of γH2AX, p-Kap-1 and p-Chk-1. Specifically Ap restored the decrease in S10 phosphorylation of histone H3 resulting from daunorubicin treatment. The cytoprotective effects of Aa and Ap were independent of daunorubicin import/export. Both Aa and Ap abrogated daunorubicin-induced accumulation of cells in S-phase. Inhibition of DNA synthesis was specific for Ap. The data show that Aa and Ap have both congruent and agent-specific pleiotropic effects that are preferential for anticancer drugs. Since Ap showed a broader spectrum of anticancer activities, this compound is suggested as novel lead compound for forthcoming in vivo studies elucidating the usefulness of spongean alkaloids in AML therapy.
Product Description
Supelco
Glycine, certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland
Supelco
Glycine, analytical standard, for nitrogen determination according to Kjeldahl method
Sigma-Aldrich
Glycine, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Glycine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
Glycine, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, ≥98.5%
Sigma-Aldrich
Glycine, BioXtra, ≥99% (titration)
SAFC
Glycine
Sigma-Aldrich
Glycine, BioUltra, Molecular Biology, ≥99.0% (NT)
Sigma-Aldrich
Glycine, tested according to Ph. Eur.
Sigma-Aldrich
Glycine, ACS reagent, ≥98.5%
Sigma-Aldrich
Glycine, 99%, FCC
Sigma-Aldrich
Glycine, puriss. p.a., reag. Ph. Eur., buffer substance, 99.7-101% (calc. to the dried substance)
Sigma-Aldrich
Glycine, meets analytical specification of Ph. Eur., BP, USP, 99-101% (based on anhydrous substance)
Supelco
Glycine, Pharmaceutical Secondary Standard; Certified Reference Material
USP
Glycine, United States Pharmacopeia (USP) Reference Standard
Glycine, European Pharmacopoeia (EP) Reference Standard