Merck
  • Wavelet-based peak detection and a new charge inference procedure for MS/MS implemented in ProteoWizard's msConvert.
Wavelet-based peak detection and a new charge inference procedure for MS/MS implemented in ProteoWizard's msConvert.
Journal of proteome research (2014-11-21)
William R French, Lisa J Zimmerman, Birgit Schilling, Bradford W Gibson, Christine A Miller, R Reid Townsend, Stacy D Sherrod, Cody R Goodwin, John A McLean, David L Tabb
ABSTRACT
We report the implementation of high-quality signal processing algorithms into ProteoWizard, an efficient, open-source software package designed for analyzing proteomics tandem mass spectrometry data. Specifically, a new wavelet-based peak-picker (CantWaiT) and a precursor charge determination algorithm (Turbocharger) have been implemented. These additions into ProteoWizard provide universal tools that are independent of vendor platform for tandem mass spectrometry analyses and have particular utility for intralaboratory studies requiring the advantages of different platforms convergent on a particular workflow or for interlaboratory investigations spanning multiple platforms. We compared results from these tools to those obtained using vendor and commercial software, finding that in all cases our algorithms resulted in a comparable number of identified peptides for simple and complex samples measured on Waters, Agilent, and AB SCIEX quadrupole time-of-flight and Thermo Q-Exactive mass spectrometers. The mass accuracy of matched precursor ions also compared favorably with vendor and commercial tools. Additionally, typical analysis runtimes (∼1-100 ms per MS/MS spectrum) were short enough to enable the practical use of these high-quality signal processing tools for large clinical and research data sets.
Sigma-Aldrich
Iodoacetic acid, ≥98.0% (T)
Sigma-Aldrich
3-Indoleacetic acid, suitable for plant cell culture, crystalline
Supelco
3-Indoleacetic acid, PESTANAL®, analytical standard
Sigma-Aldrich
3-Indoleacetic acid, 98%
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
N,N-Dimethyldodecylamine, 97%
Sigma-Aldrich
Indole-3-acetic acid sodium salt, suitable for plant cell culture, BioReagent, ≥98%