Merck
  • Influence of polymer end-chemistry on the morphology of perfluorohexane polymeric microcapsules intended as ultrasound contrast agents.
Influence of polymer end-chemistry on the morphology of perfluorohexane polymeric microcapsules intended as ultrasound contrast agents.
International journal of pharmaceutics (2014-05-20)
L Mousnier, N Huang, E Morvan, E Fattal, N Tsapis
ABSTRACT
Ultrasound contrast agents (UCAs) composed of a liquid perfluorocarbon (PFC) core surrounded by a polymer shell have shown promising echogenicity as well as stability. In a strategy to optimize the ultrasound properties of these systems, encapsulating a liquid PFC with a low boiling point such as perfluorohexane (PFH) was suggested. The ultimate aim of these systems would be to induce phase-transition of the liquid PFH into gas by acoustic droplet vaporization (ADV) to further increase the UCA acoustic response. Microcapsules with a perfluorohexane core have been developed by an emulsion-evaporation process, using three biodegradable polymers: PLGA and PLA with acid (PLA-COOH) and ester (PLA-COOR) terminations. Despite their similar properties, these polymers were found to strongly influence the final microcapsule morphology. While PLGA was able to form nice core-shell microcapsules, the use of PLA-COOH led to decentered microcapsules and big "eye" morphologies, and PLA-COOR induced the formation of "acorn" morphologies. To shed light on morphologies disparities, polymer interfacial behavior was studied at the dichloromethane-water and the PFH-dichloromethane interfaces. One can conclude that the core-shell structure is the result of a significant adsorption of the polymer at the dichloromethane-water interface together with a good stability of the PFH droplet within the emulsion globule. Previous work has shown that the capsule's thickness-to-radius (T/R) ratio can be controlled easily by varying the polymer to perfluorocarbon proportions. This versatility was confirmed for PFH capsules with PLA-COOH and PLGA shells. Finally, the encapsulation efficiency of PFH was assessed by relating the T/R ratio to the volume fraction of PFH and by (19)F NMR spectroscopy.
Sigma-Aldrich
Tetradecafluorohexane, 99%
Sigma-Aldrich
Tetradecafluorohexane, A mixture of perfluorinated hexanes, 95%
Supelco
Dichloromethane, analytical standard
Supelco
Dichloromethane, Selectophore, ≥99.5%
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.9%, contains 40-150 ppm amylene as stabilizer
Supelco
Methylene Chloride, Pharmaceutical Secondary Standard; Certified Reference Material
Sigma-Aldrich
Dichloromethane, contains 40-150 ppm amylene as stabilizer, ACS reagent, ≥99.5%
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, suitable for HPLC, ≥99.8%, contains amylene as stabilizer
Sigma-Aldrich
Dichloromethane, HPLC Plus, for HPLC, GC, and residue analysis, ≥99.9%, contains 50-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, ACS reagent, ≥99.5%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Dichloromethane, biotech. grade, 99.9%, contains 40-150 ppm amylene as stabilizer
Sigma-Aldrich
Zirconyl chloride octahydrate, reagent grade, 98%
Sigma-Aldrich
Diethyl ether
Supelco
Diethyl ether, analytical standard
Sigma-Aldrich
Diethyl ether, anhydrous, ACS reagent, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, suitable for HPLC, ≥99.9%, inhibitor-free
Sigma-Aldrich
Diethyl ether, ACS reagent, anhydrous, ≥99.0%, contains BHT as inhibitor
Sigma-Aldrich
Diethyl ether, ACS reagent, ≥98.0%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor
Sigma-Aldrich
Diethyl ether, reagent grade, ≥98%, contains ≤2% ethanol and ≤10ppm BHT as inhibitor