Merck
  • Structures of the reaction products of tetraalkoxytitanium with acetylacetone and ethyl acetoacetate.
Structures of the reaction products of tetraalkoxytitanium with acetylacetone and ethyl acetoacetate.
Journal of the American Chemical Society (1957)
Yamamoto A and Kambara .