Merck
  • CD95 rs1800682A/G variant and tumor risk in Asians: evidence from a meta-analysis of 36 case-control studies containing 22,438 samples.
CD95 rs1800682A/G variant and tumor risk in Asians: evidence from a meta-analysis of 36 case-control studies containing 22,438 samples.
Medical science monitor : international medical journal of experimental and clinical research (2015-02-28)
Cheng Jin, Xiaomin Wu, Yuanlong Gu, Fenglai Yuan, Qinghai Ye, Feng Dai, Lijie Zhu, Yuanyuan Mi
ABSTRACT
The CD95 gene plays a key role in regulating cell growth and tumor genesis. To date, several publications have focused on the CD95 rs1800682A/G site polymorphism and various types of tumors in Asians; however, this association is still controversial and obscure. Therefore, a meta-analysis combined with all publications to clarify this association is necessary. A search in the PubMed and SinoMed databases was performed to detect all relevant included publications. Odds ratio (OR) and 95% confidence intervals (CI) revealed association strengths. Overall, 36 case-control studies were chosen based on the search criteria. There was no association of the CD95 rs1800682A/G site polymorphism with tumor risk in total and ethnicity subgroup analysis. However, further stratified analysis in the cancer subgroup revealed weakly significant associations in hepatocellular carcinoma (AA+AG vs. GG: OR=0.93, 95% CI=0.87-0.99, P=0.035; AG vs. GG: OR=0.89, 95% CI=0.80-0.99, P=0.036). The CD95 rs1800682A/G site polymorphism may be associated with hepatocellular carcinoma susceptibility. Further large-scale and well-designed studies regarding tumor types and ethnicities are still required to confirm our results.
Sigma-Aldrich
Fas Intracellular Domain human, >90% (SDS-PAGE), recombinant, expressed in E. coli (N-terminal histidine tagged), buffered aqueous solution
Pricing and availability is not currently available.
Sigma-Aldrich
Fas/Fc Chimera from mouse, >97% (SDS-PAGE), recombinant, expressed in baculovirus infected Sf21 cells, lyophilized powder, suitable for cell culture
Pricing and availability is not currently available.