Merck
  • Modulation of mature cystic fibrosis transmembrane regulator protein by the PDZ domain protein CAL.
Modulation of mature cystic fibrosis transmembrane regulator protein by the PDZ domain protein CAL.
The Journal of biological chemistry (2003-10-23)
Jie Cheng, Hua Wang, William B Guggino
ABSTRACT
We have previously identified the cystic fibrosis transmembrane regulator (CFTR)-interacting protein CAL and demonstrated that CAL modulates CFTR plasma membrane expression by retaining CFTR within the cell. Here, we report that in addition to regulating membrane expression, CAL also regulates the expression of mature CFTR. The co-expression of hemagglutinin-tagged or Myc-tagged CAL with green fluorescent protein (GFP)-CFTR in COS-7 cells causes a dose-dependent reduction in mature GFP-CFTR, independent of its tags. Bafilomycin A1, a lysosomal proton pump inhibitor, increases mature GFP-CFTR, confirming previous reports of lysosomal degradation of mature CFTR. Importantly, bafilomycin A1 reverses CAL-mediated CFTR degradation. The proteasome inhibitor, MG132, on the other hand, does not reverse the effect of CAL. CAL has no effect on CFTR maturation, suggesting that it exerts its effects on mature CFTR. Co-expression of CAL enhances the degradation of CFTR. We showed previously that CAL reduces the half-life of CFTR at the cell surface. Here we show that expression of dominant-negative dynamin 2 K44A, a large GTPase inhibitor that is known to inhibit clathrin-mediated endocytosis and vesicle formation in the Golgi, increases cell surface CFTR as measured by surface biotinylation. More importantly, dynamin 2 K44A also restores cell surface CFTR in CAL-overexpressing cells and partially blocks the CAL-mediated degradation of mature CFTR. These data suggest a model in which CAL retains CFTR in the cell and targets CFTR for degradation.
Sigma-Aldrich
Anti-CFTR Antibody, a.a. 1370-1380, clone M3A7, clone M3A7, Chemicon®, from mouse