Merck
  • EphB2 regulates contact-dependent and contact-independent signaling to control platelet function.
EphB2 regulates contact-dependent and contact-independent signaling to control platelet function.
Blood (2014-11-06)
Sakthivel Vaiyapuri, Tanya Sage, Rekha H Rana, Michael P Schenk, Marfoua S Ali, Amanda J Unsworth, Chris I Jones, Alexander R Stainer, Neline Kriek, Leonardo A Moraes, Jonathan M Gibbins
ABSTRACT
The Eph kinases, EphA4 and EphB1, and their ligand, ephrinB1, have been previously reported to be present in platelets where they contribute to thrombus stability. Although thrombus formation allows for Eph-ephrin engagement and bidirectional signaling, the importance specifically of Eph kinase or ephrin signaling in regulating platelet function remained unidentified. In the present study, a genetic approach was used in mice to establish the contribution of signaling orchestrated by the cytoplasmic domain of EphB2 (a newly discovered Eph kinase in platelets) in platelet activation and thrombus formation. We conclude that EphB2 signaling is involved in the regulation of thrombus formation and clot retraction. Furthermore, the cytoplasmic tail of this Eph kinase regulates initial platelet activation in a contact-independent manner in the absence of Eph-ephrin ligation between platelets. Together, these data demonstrate that EphB2 signaling not only modulates platelet function within a thrombus but is also involved in the regulation of the function of isolated platelets in a contact-independent manner.
Sigma-Aldrich
EphB2/Fc Chimera from mouse, >90% (SDS-PAGE), recombinant, expressed in NSO cells, lyophilized powder
Pricing and availability is not currently available.