- Pd-catalyzed multidehydrogenative cross-coupling between (hetero)arenes and nitroethane to construct β-aryl nitroethylenes.
Pd-catalyzed multidehydrogenative cross-coupling between (hetero)arenes and nitroethane to construct β-aryl nitroethylenes.
Organic letters (2013-03-27)
Min Zhang, Peng Hu, Jun Zhou, Ge Wu, Shijun Huang, Weiping Su
PMID23528043
ABSTRACT
The Pd-catalyzed multidehydrogenative cross-coupling reactions of arenes with nitroethane are described. The established methods afford β-aryl nitroethylenes that are an important class of synthetic intermediates and biologically active compounds in an atom- and step-economical fashion. The reactions were applicable to substituted benzenes and a variety of heterocycles such as benzothiophenes, benzofurans, and indoles. Mechanistic experiments indicated that β-nitroethylbenzene might be the intermediate in this transformation.