Merck
  • Unsupervised explorative data analysis of normal human leukocytes and BCR/ABL positive leukemic cells mid-infrared spectra.
Unsupervised explorative data analysis of normal human leukocytes and BCR/ABL positive leukemic cells mid-infrared spectra.
The Analyst (2015-05-20)
G Bellisola, M Bolomini Vittori, G Cinque, P Dumas, Z Fiorini, C Laudanna, M Mirenda, C Sandt, G Silvestri, L Tomasello, M Vezzalini, K Wehbe, C Sorio
ABSTRACT
We proved the ability of Fourier Transform Infrared microspectroscopy (microFTIR) complemented by Principal Component Analysis (PCA) to detect protein phosphorylation/de-phosphorylation in mammalian cells. We analyzed by microFTIR human polymorphonuclear neutrophil (PMNs) leukocytes, mouse-derived parental Ba/F3 cells (Ba/F3#PAR), Ba/F3 cells transfected with p210(BCR/ABL) (Ba/F3#WT) and expressing high levels of protein tyrosine kinase (PTK), and human-derived BCR/ABL positive K562 leukemic cell sub-clones engineered to differently express receptor-type tyrosine-protein phosphatase gamma (PTPRG). Synchrotron radiation (SR) and conventional (globar) IR sources were used to perform microFTIR respectively, on single cells and over several cells within the same sample. Ex vivo time-course experiments were run, inducing maximal protein phosphorylation in PMNs by 100 nM N-formylated tripeptide fMLP. Within the specific IR fingerprint 1800-850 cm(-1) frequency domain, PCA identified two regions with maximal signal variance. These were used to model and test the robustness of PCA in representing the dynamics of protein phosphorylation/de-phosphorylation processes. An IR signal ratio marker reflecting the homeostatic control by protein kinases and phosphatases was identified in normal leukocytes. The models identified by microFTIR and PCA in normal leukocytes also distinguished BCR/ABL positive Ba/F3#WT from BCR/ABL negative Ba/F3#PAR cells as well as K562 cells exposed to functionally active protein tyrosine phosphatase recombinant protein ICD-Tat transduced in cells by HIV-1 Tat technology or cells treated with the PTK inhibitor imatinib mesylate (IMA) from cells exposed to phosphatase inactive (D1028A)ICD-Tat recombinant protein and untreated control cells, respectively. The IR signal marker correctly reflected the degrees of protein phosphorylation associated with abnormal PTK activity in BCR/ABL positive leukemic cells and in general was inversely related to the expression/activity of PTPRG in leukemic sub-clones. In conclusion, we have described a new, reliable and simple spectroscopic method to study the ex vivo protein phosphorylation/de-phosphorylation balance in cell models: it is suitable for biomedical and pharmacological research labs but it also needs further optimization and its evaluation on large cohorts of patients to be proposed in the clinical setting of leukemia.
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~0.025 M in H2O
Sigma-Aldrich
N,N′-Methylenebis(acrylamide), 99%
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, powder, BioReagent, suitable for cell culture, ≥98% (HPLC), suitable for fluorescence
Sigma-Aldrich
Ammonium molybdate tetrahydrate, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Magnesium chloride solution, PCR Reagent, 25 mM MgCI2 solution for PCR
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioReagent, suitable for cell culture, suitable for insect cell culture, 81.0-83.0% MoO3 basis
Sigma-Aldrich
Magnesium chloride solution, 0.1 M
Sigma-Aldrich
Magnesium chloride solution, Molecular Biology, 1.00 M±0.01 M
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, ~1 M in H2O
Sigma-Aldrich
Magnesium chloride solution, BioUltra, Molecular Biology, 2 M in H2O
Sigma-Aldrich
N,N′-Methylenebisacrylamide, suitable for electrophoresis (after filtration or allowing insolubles to settle)
Sigma-Aldrich
Lauric acid, natural, ≥98%, FCC, FG
Sigma-Aldrich
Acrylamide, Molecular Biology, ≥99% (HPLC)
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99%
Sigma-Aldrich
Acrylamide, suitable for electrophoresis, ≥99% (HPLC), powder
Sigma-Aldrich
4′,6-Diamidino-2-phenylindole dihydrochloride, suitable for fluorescence, BioReagent, ≥95.0% (HPLC)
Sigma-Aldrich
Ammonium molybdate tetrahydrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Ammonium molybdate, 99.98% trace metals basis
Sigma-Aldrich
Ammonium molybdate, meets USP testing specifications
Sigma-Aldrich
Lauric acid, ≥98%, FCC, FG
Sigma-Aldrich
N,N′-Methylenebisacrylamide, powder, Molecular Biology, suitable for electrophoresis, ≥99.5%
Sigma-Aldrich
DL-Tyrosine, 99%
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, BioXtra, ≥99.0% (TLC)
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
N,N′-Methylenebisacrylamide solution, suitable for electrophoresis, 2% in H2O
Sigma-Aldrich
Acrylamide solution, 40%, suitable for electrophoresis, sterile-filtered
Sigma-Aldrich
N-Formyl-Met-Leu-Phe, ≥97% (HPLC)
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.99% trace metals basis
Sigma-Aldrich
Magnesium chloride, powder, <200 μm
Sigma-Aldrich
Magnesium chloride, AnhydroBeads, −10 mesh, 99.9% trace metals basis