Merck
  • New valid spectrofluorimetric method for determination of selected cephalosporins in different pharmaceutical formulations using safranin as fluorophore.
New valid spectrofluorimetric method for determination of selected cephalosporins in different pharmaceutical formulations using safranin as fluorophore.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy (2015-10-12)
Sayed M Derayea, Hytham M Ahmed, Osama H Abdelmageed, Ahmed M Haredy
ABSTRACT
A new validated spectrofluorimetric method has been developed for the determination of some cephalosporins namely; cefepime, cefaclor, cefadroxil, cefpodoxime and cefexime. The method was based on the reaction of these drugs with safranin in slightly alkaline medium (pH 8.0), to form ion-association complexes. The fluorescent products were extracted into chloroform and their fluorescence intensities were measured at 544-565 nm after excitation at 518-524 nm. The reaction conditions influencing the product formation and stability were investigated and optimized. The relative fluorescence intensity was proportional to the drug concentration in the linear ranges of 0.15-1.35, 0.35-1.25, 0.35-1.25, 0.20-1.44 and 0.20-1.25 μg/mL for cefepime, cefaclor, cefadroxil, cefpodoxime proxetil and cefexime, respectively. The detection limits were 40, 100, 100, 60 and 70 ng/mL, respectively. The performance of the developed method was evaluated in terms of Student's t-test and variance ratio F-test to find out the significance of proposed methods over the reference spectrophotometric method. Various pharmaceutical formulations were successfully analyzed using the proposed method and the results were in good agreement with those of the previously reported methods.
Sigma-Aldrich
Cefaclor
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in acetic acid
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrogen chloride solution, 4.0 M in dioxane
Sigma-Aldrich
Hydrochloric acid, meets analytical specification of Ph. Eur., BP, NF, fuming, 36.5-38%
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid, ACS reagent, 37%
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Hydrochloric acid, 37 wt. % in H2O, 99.999% trace metals basis
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Hydrogen chloride solution, 1.0 M in diethyl ether
Sigma-Aldrich
Hydrogen chloride solution, 2.0 M in diethyl ether