Merck
  • Targeting the Spleen Tyrosine Kinase with Fostamatinib as a Strategy against Waldenström Macroglobulinemia.
Targeting the Spleen Tyrosine Kinase with Fostamatinib as a Strategy against Waldenström Macroglobulinemia.
Clinical cancer research : an official journal of the American Association for Cancer Research (2015-03-10)
Isere Kuiatse, Veerabhadran Baladandayuthapani, Heather Y Lin, Sheeba K Thomas, Chad C Bjorklund, Donna M Weber, Michael Wang, Jatin J Shah, Xing-Ding Zhang, Richard J Jones, Stephen M Ansell, Guang Yang, Steven P Treon, Robert Z Orlowski
ABSTRACT
Waldenström macroglobulinemia (WMG) is a lymphoproliferative disorder characterized by good initial responses to standard therapeutics, but only a minority of patients achieve complete remissions, and most inevitably relapse, indicating a need for novel agents. B-cell receptor signaling has been linked to clonal evolution in WMG, and Spleen tyrosine kinase (Syk) is overexpressed in primary cells, suggesting that it could be a novel and rational target. We studied the impact of the Syk inhibitor fostamatinib on BCWM.1 and MWCL-1 WMG-derived cell lines both in vitro and in vivo, as well as on primary patient cells. In WMG-derived cell lines, fostamatinib induced a time- and dose-dependent reduction in viability, associated with activation of apoptosis. At the molecular level, fostamatinib reduced activation of Syk and Bruton's tyrosine kinase, and also downstream signaling through MAPK kinase (MEK), p44/42 MAPK, and protein kinase B/Akt. As a single agent, fostamatinib induced tumor growth delay in an in vivo model of WMG, and reduced viability of primary WMG cells, along with inhibition of p44/42 MAPK signaling. Finally, fostamatinib in combination with other agents, including dexamethasone, bortezomib, and rituximab, showed enhanced activity. Taken together, these data support the translation of approaches targeting Syk with fostamatinib to the clinic for patients with relapsed and possibly even newly diagnosed WMG.
Sigma-Aldrich
Anti-β-Actin antibody, Mouse monoclonal, clone AC-15, purified from hybridoma cell culture
Sigma-Aldrich
Propidium iodide solution
Sigma-Aldrich
Dimethyl sulfide, ≥99%
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, BioUltra, ≥99.5% (NT)
Sigma-Aldrich
Dimethyl sulfide, ≥99%, FCC, FG
Sigma-Aldrich
Dimethyl sulfide, anhydrous, ≥99.0%
Sigma-Aldrich
Dexamethasone, powder, BioReagent, suitable for cell culture, ≥97%
Sigma-Aldrich
Dexamethasone, powder, γ-irradiated, BioXtra, suitable for cell culture, ≥80% (HPLC)
Sigma-Aldrich
2-Butanone, FCC, FG
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
Dexamethasone, ≥98% (HPLC), powder
Sigma-Aldrich
L-Glutamine
SAFC
L-Glutamine
Sigma-Aldrich
Dimethyl sulfide, natural, ≥99%, FCC, FG
Sigma-Aldrich
Dexamethasone, meets USP testing specifications
Sigma-Aldrich
meso-2,3-Dimercaptosuccinic acid, ~98%
Sigma-Aldrich
Propidium iodide, ≥94.0% (HPLC)