Merck
  • Nitric oxide and oxidative stress is associated with severity of diabetic retinopathy and retinal structural alterations.
Nitric oxide and oxidative stress is associated with severity of diabetic retinopathy and retinal structural alterations.
Clinical & experimental ophthalmology (2015-02-14)
Shashi Sharma, Sandeep Saxena, Khushboo Srivastav, Rajendra K Shukla, Nibha Mishra, Carsten H Meyer, Peter Kruzliak, Vinay K Khanna
ABSTRACT
The aim of the study was to determine plasma nitric oxide (NO) and lipid peroxide (LPO) levels in diabetic retinopathy and its association with severity of disease. Prospective observational study. A total of 60 consecutive cases and 20 healthy controls were included. Severity of retinopathy was graded according to early treatment diabetic retinopathy study (ETDRS) classification. Photoreceptor inner segment ellipsoid band (ISel) disruption and retinal pigment epithelium (RPE) alteration were graded using spectral domain optical coherence tomography. Data were statistically analyzed. Plasma thiobarbituric acid reactive substances, NO assay and reduced glutathione (GSH) were measured using standard protocol. Increased severity of diabetic retinopathy was significantly associated with increase in plasma levels of LPO (P < 0.05), NO (P < 0.001) and decrease in plasma levels of GSH (P < 0.0001), ISel disruption (P < 0.001) and RPE topographic alteration (P < 0.01). Increased plasma NO levels are associated with increased severity of diabetic retinopathy. For the first time, it has been demonstrated that increased plasma LPO, NO and decreased GSH levels are associated with in vivo structural changes in inner segment ellipsoid and RPE.
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Sodium trichloroacetate, 97%
Sigma-Aldrich
Trichloroacetic acid, BioXtra, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, ≥99.0%
Sigma-Aldrich
Trichloroacetic acid, ≥99.0% (titration)
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ≥98%, BioReagent, suitable for determination of sulfhydryl groups
Sigma-Aldrich
Trichloroacetic acid, suitable for electrophoresis, suitable for fixing solution (for IEF and PAGE gels), ≥99%
Sigma-Aldrich
Trichloroacetic acid, BioUltra, ≥99.5% (T)
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
5,5′-Dithiobis(2-nitrobenzoic acid), ReagentPlus®, 99%
Sigma-Aldrich
Trichloroacetic acid, ACS reagent, for the determination of Fe in blood according to Heilmeyer, ≥99.5%
Sigma-Aldrich
Benzoic acid, purified by sublimation, ≥99%
Sigma-Aldrich
Trichloroacetic acid solution, 6.1 N
Sigma-Aldrich
Ethylenediaminetetraacetic acid, 99.995% trace metals basis
Sigma-Aldrich
Benzoic acid, ACS reagent, ≥99.5%
Sigma-Aldrich
Benzoic acid, ReagentPlus®, 99%
Sigma-Aldrich
Benzoic acid, natural, ≥99.5%, FCC, FG
Sigma-Aldrich
Ethylenediamine, ReagentPlus®, ≥99%
Sigma-Aldrich
Benzoic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Benzoic acid, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99.9% (alkalimetric)
Sigma-Aldrich
Benzoic acid, meets analytical specification of Ph. Eur., BP, USP, FCC, E210, 99.5-100.5% (alkalimetric)
Sigma-Aldrich
Ethylenediamine solution, technical, 75-80%
Sigma-Aldrich
Ethylenediamine, purified by redistillation, ≥99.5%
Sigma-Aldrich
Ethylenediaminetetraacetic acid, anhydrous, crystalline, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylenediaminetetraacetic acid, BioUltra, anhydrous, ≥99% (titration)
Sigma-Aldrich
Ethylenediaminetetraacetic acid, purified grade, ≥98.5%, powder
Sigma-Aldrich
Ethylenediaminetetraacetic acid, ACS reagent, 99.4-100.6%, powder