Merck
  • Dual-Wavelength Irradiation and Dox Delivery for -Cancer Cell Ablation with Photocatalytic Pr Doped TiO2 /NGO -Hybrid Nanocomposite.
Dual-Wavelength Irradiation and Dox Delivery for -Cancer Cell Ablation with Photocatalytic Pr Doped TiO2 /NGO -Hybrid Nanocomposite.
Advanced healthcare materials (2015-06-19)
Hongje Jang, Myung-Ho Choi, Yeajee Yim, Young-Kwan Kim, Dal-Hee Min
ABSTRACT
Herein, hybrid nanocomposite of praseodymium doped TiO2 nanocrystals and graphene oxide nanosheets are prepared by facile hydrothermal treatment. As-synthesized Pr-TiO2 /NGO hybrid nanocomposite exhibits enhanced photocatalytic activity under visible light irradiation by the intact graphene oxide and doped lanthanide mediated band gap narrowing compared to TiO2 . Moreover, high payload and controlled release of doxorubicin by charge reversal of hybrid nanocomposite at endosomal pH and near-infrared irradiation mediated efficient photothermal conversion provide highly favorable features in therapeutic applications. Through the combination of these three distinctive therapeutic modalities, highly efficient trimodal cancer cell ablation is demonstrated.
Sigma-Aldrich
Titanium(IV) butoxide, reagent grade, 97%
Sigma-Aldrich
Ethylene glycol, anhydrous, 99.8%
Sigma-Aldrich
Sodium nitrate-14N, 99.95 atom % 14N
Sigma-Aldrich
Hydrogen peroxide solution, 30 % (w/w) in H2O, contains stabilizer
Sigma-Aldrich
Sodium nitrate, ≥99.0%, suitable for plant cell culture
Sigma-Aldrich
Sodium nitrate, BioXtra, ≥99.0%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
Doxorubicin hydrochloride, 98.0-102.0% (HPLC)
Sigma-Aldrich
Sodium nitrate, 99.995% trace metals basis
Sigma-Aldrich
Hydrogen peroxide solution, 34.5-36.5%
Sigma-Aldrich
Sodium nitrate, BioUltra, ≥99.0% (T)
Sigma-Aldrich
Doxorubicin hydrochloride, suitable for fluorescence, 98.0-102.0% (HPLC)
Sigma-Aldrich
Ethylene glycol, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Diethyl azodicarboxylate solution, purum, ~40% in toluene (H-NMR)
Supelco
Ethylene glycol solution, suitable for NMR (reference standard), 80% in DMSO-d6 (99.9 atom % D), NMR tube size 5 mm × 8 in.
Sigma-Aldrich
Ethylene glycol 5 M solution