Merck
  • Iodine status of young Burkinabe children receiving small-quantity lipid-based nutrient supplements and iodised salt: a cluster-randomised trial.
Iodine status of young Burkinabe children receiving small-quantity lipid-based nutrient supplements and iodised salt: a cluster-randomised trial.
The British journal of nutrition (2015-09-29)
Sonja Y Hess, Souheila Abbeddou, Elizabeth Yakes Jimenez, Jean-Bosco Ouédraogo, Kenneth H Brown
ABSTRACT
The objective of the present study was to assess the impact of providing small-quantity lipid-based nutrient supplements (SQ-LNS) on the I status of young Burkinabe children. In total, thirty-four communities were assigned to intervention (IC) or non-intervention cohorts (NIC). IC children were randomly assigned to receive 20 g lipid-based nutrient supplements (LNS)/d containing 90 µg I with 0 or 10 mg Zn from 9 to 18 months of age, and NIC children received no SQ-LNS. All the children were exposed to iodised salt through the national salt iodization programme. Spot urinary iodine (UI), thyroid-stimulating hormone (TSH) and total thyroxine (T4) in dried blood spots as well as plasma thyroglobulin (Tg) concentrations were assessed at 9 and 18 months of age among 123 IC and fifty-six NIC children. At baseline and at 18 months, UI, TSH and T4 did not differ between cohorts. Tg concentration was higher in the NIC v. IC at baseline, but this difference did not persist at 18 months of age. In both cohorts combined, the geometric mean of UI was 339·2 (95% CI 298·6, 385·2) µg/l, TSH 0·8 (95% CI 0·7, 0·8) mU/l, T4 118 (95 % CI 114, 122) nmol/l and Tg 26·0 (95% CI 24·3, 27·7) µg/l at 18 months of age. None of the children had elevated TSH at 18 months of age. Marginally more children in NIC (8·9%) had low T4 (15 ppm). A reduction of SQ-LNS I content could be considered in settings with similarly successful salt iodisation programmes.
Sigma-Aldrich
L-Lysine monohydrochloride, from non-animal source, meets EP, JP, USP testing specifications, suitable for cell culture, 98.5-101.0%
Sigma-Aldrich
L-Lysine monohydrochloride, BioUltra, ≥99.5% (AT)
Sigma-Aldrich
L-Lysine monohydrochloride, reagent grade, ≥98% (HPLC)
Sigma-Aldrich
Iodine, ReagentPlus®, 99.7% trace metals basis, beads, 1-3 mm
Sigma-Aldrich
Iodine, ACS reagent, ≥99.8%, solid
Supelco
Iodine, ReagentPlus®, ≥99.8% (titration)
Sigma-Aldrich
Iodine, flakes, ReagentPlus®, ≥99%
Sigma-Aldrich
Iodine, ≥99.99% trace metals basis
Sigma-Aldrich
Iodine, 99.999% trace metals basis
Sigma-Aldrich
Iodine, anhydrous, beads, −10 mesh, 99.999% trace metals basis