Merck
  • Immunomodulatory activity on macrophage of a purified polysaccharide extracted from Laminaria japonica.
Immunomodulatory activity on macrophage of a purified polysaccharide extracted from Laminaria japonica.
Carbohydrate polymers (2015-10-03)
Qing Fang, Jing-Fei Wang, Xue-Qiang Zha, Shao-Hua Cui, Lei Cao, Jian-Ping Luo
ABSTRACT
In this work, a novel water-soluble homogeneous polysaccharide (LJP-31) with a molecular mass of 2.24 × 10(6) Da was isolated and purified from Laminaria japonica using DEAE-cellulose and Sephacryl S500 chromatography. Results showed that LJP-31 mainly consists of arabinose, mannose, glucose and galactose in a molar ratio of 1.0:7.8:6.6:0.8. LJP-31 exhibited significant stimulation on macrophages and enhanced the production of NO, TNF-α, IL-1β, IL-6 and IL-10 as well as the up-regulation of their gene expressions. Western blot analysis suggested that LJP-31 has the positive effects on the translocation of NF-κB p65 from cytoplasm to nucleus and the phosphorylation of IκBα, ERK1/2, JNK1/2 and P38 in macrophages. Flow cytometric and confocal laser-scanning microscopy analysis indicated that toll-like receptor 4 (TLR4) was at least one of the recognition receptors of LJP-31 on the plasma membrane of macrophages. Taken together, LJP-31 may exert its immunostimulating potency via TLR4 activation of MAPK and NF-κB signaling pathways.
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, ReagentPlus®, ≥99.5%
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, ACS reagent, ≥99.9%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Xylose, ≥99%
Sigma-Aldrich
D-(+)-Xylose, BioXtra, ≥99% (GC)
Sigma-Aldrich
D-(+)-Xylose, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Xylose, ≥99% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, 98%
Sigma-Aldrich
Thiazolyl Blue Tetrazolium Bromide, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥97.5% (HPLC)
Sigma-Aldrich
L-(+)-Arabinose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Mannose, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
D-(+)-Mannose, BioUltra, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-(+)-Mannose, ≥99% (GC), wood
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Galactose, ≥99% (HPLC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Galactose, ≥98% (HPLC)