Merck
  • Impact of vitamin C on the cardiometabolic and inflammatory profiles of mice lacking a functional Werner syndrome protein helicase.
Impact of vitamin C on the cardiometabolic and inflammatory profiles of mice lacking a functional Werner syndrome protein helicase.
Experimental gerontology (2015-11-03)
Lucie Aumailley, Marie Julie Dubois, Chantal Garand, André Marette, Michel Lebel
ABSTRACT
Werner syndrome (WS) is a premature aging disorder caused by mutations in a DNA helicase/exonuclease. Mice lacking the helicase domain of this protein exhibit metabolic abnormalities that are reversed by vitamin C. In this study, we used a targeted metabolomic approach to identify serum metabolites significantly altered in young mutant mice treated with or without vitamin C. We also measured several serum inflammatory and cardiometabolic factors. We show that young mutant mice exhibit an increase in serum hydroxyproline and plasminogen activator inhibitor-1 (PAI-1), markers of cardiovascular diseases and inflammation, before they exhibit morphological anomalies in different tissues. We also observed an increase in three very long chain lysophosphatidylcholines underlying peroxisome perturbation. Vitamin C reversed the concentrations of these metabolites and PAI-1 to wild type values. Transcriptomic analyses on the liver of mutant mice revealed a decrease in the expression of genes involved in fatty acid degradation compared to wild type animals. Vitamin C treatment increased the expression of genes involved in glutathione metabolism and the synthesis of unsaturated fatty acids in these mice. These results show that changes at the transcriptomic level concord with the alterations of several serum metabolites in these mice. Finally, we found that a mislocalization of the Wrn mutant protein in the liver endoplasmic reticulum fraction increased oxidative stress in that cellular compartment. Vitamin C reversed this oxidative stress. To conclude, this study provides novel potential predictive cardiometabolic biomarkers in WS that will allow the assessment of the impact of vitamin C on patients with WS.
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Supelco
L-Ascorbic acid, analytical standard
Sigma-Aldrich
Retinol, synthetic, ≥95% (HPLC), (Powder or Powder with Lumps)
Sigma-Aldrich
Retinol, BioXtra, ≥97.5% (HPLC), ~3100 U/mg
Sigma-Aldrich
Retinol, ≥95.0% (HPLC), ~2700 U/mg
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium pyrophosphate tetrabasic, ≥95%
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, Molecular Biology, ≥98.5% (GC), free-flowing, Redi-Dri
Sigma-Aldrich
Sodium dodecyl sulfate, ≥98.0% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, BioXtra, ≥99.0% (GC)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium Fluoride Solution
Sigma-Aldrich
Sodium orthovanadate, ≥90% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, 92.5-100.5% based on total alkyl sulfate content basis
Supelco
Sodium dodecyl sulfate, dust-free pellets, suitable for electrophoresis, Molecular Biology, ≥99.0% (GC)
Sigma-Aldrich
Sodium orthovanadate, 99.98% trace metals basis
Sigma-Aldrich
Sodium dodecyl sulfate, BioReagent, suitable for electrophoresis, Molecular Biology, ≥98.5% (GC)
Sigma-Aldrich
Sodium dodecyl sulfate, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium dodecyl sulfate, ≥99.0% (GC), dust-free pellets
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium dodecyl sulfate, ≥90% ((Assay))
Sigma-Aldrich
Sodium dodecyl sulfate, tested according to NF, mixture of sodium alkyl sulfates consisting mainly of sodium dodecyl sulfate
Sigma-Aldrich
Sodium dodecyl sulfate, BioUltra, Molecular Biology, ≥99.0% (GC)