Merck
Mutational Analysis of the Chlamydia muridarum Plasticity Zone.
Infection and immunity (2015-05-06)
Krithika Rajaram, Amanda M Giebel, Evelyn Toh, Shuai Hu, Jasmine H Newman, Sandra G Morrison, Laszlo Kari, Richard P Morrison, David E Nelson
ABSTRACT
Pathogenically diverse Chlamydia spp. can have surprisingly similar genomes. Chlamydia trachomatis isolates that cause trachoma, sexually transmitted genital tract infections (chlamydia), and invasive lymphogranuloma venereum (LGV) and the murine strain Chlamydia muridarum share 99% of their gene content. A region of high genomic diversity between Chlamydia spp. termed the plasticity zone (PZ) may encode niche-specific virulence determinants that dictate pathogenic diversity. We hypothesized that PZ genes might mediate the greater virulence and gamma interferon (IFN-γ) resistance of C. muridarum compared to C. trachomatis in the murine genital tract. To test this hypothesis, we isolated and characterized a series of C. muridarum PZ nonsense mutants. Strains with nonsense mutations in chlamydial cytotoxins, guaBA-add, and a phospholipase D homolog developed normally in cell culture. Two of the cytotoxin mutants were less cytotoxic than the wild type, suggesting that the cytotoxins may be functional. However, none of the PZ nonsense mutants exhibited increased IFN-γ sensitivity in cell culture or were profoundly attenuated in a murine genital tract infection model. Our results suggest that C. muridarum PZ genes are transcribed--and some may produce functional proteins--but are dispensable for infection of the murine genital tract.
Sigma-Aldrich
Magnesium sulfate solution, BioUltra, Molecular Biology
Sigma-Aldrich
Sodium carbonate-12C, 99.9 atom % 12C
Sigma-Aldrich
Magnesium sulfate, ≥99.99% trace metals basis
Sigma-Aldrich
Ethyl methanesulfonate, liquid
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Magnesium sulfate, BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Magnesium sulfate solution, Molecular Biology, 1.00 M±0.04 M
Sigma-Aldrich
Sodium carbonate, anhydrous, powder, 99.999% trace metals basis
Sigma-Aldrich
Sodium carbonate, BioUltra, anhydrous, ≥99.5% (calc. on dry substance, T)
Sigma-Aldrich
1,1,2,2-Tetrabromoethane, 98%
Sigma-Aldrich
Sodium pyruvate, BioXtra, ≥99%
Sigma-Aldrich
Sodium pyruvate, ReagentPlus®, ≥99%
Sigma-Aldrich
Sodium carbonate, BioXtra, ≥99.0%
Sigma-Aldrich
Sodium pyruvate, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, ≥99%
Sigma-Aldrich
Sodium pyruvate, powder, BioXtra, suitable for mouse embryo cell culture
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥98.5% (GC)
Sigma-Aldrich
Sodium pyruvate, Hybri-Max, powder, suitable for hybridoma
Sigma-Aldrich
Phenylmethanesulfonyl fluoride, ≥99.0% (T)
Sigma-Aldrich
Sodium pyruvate, anhydrous, free-flowing, Redi-Dri, ReagentPlus®, ≥99%