Merck
  • Metallothionein, essential elements and lipid peroxidation in mercury-exposed suckling rats pretreated with selenium.
Metallothionein, essential elements and lipid peroxidation in mercury-exposed suckling rats pretreated with selenium.
Biometals : an international journal on the role of metal ions in biology, biochemistry, and medicine (2015-05-08)
Tatjana Orct, Maja Lazarus, Marija Ljubojević, Ankica Sekovanić, Ivan Sabolić, Maja Blanuša
ABSTRACT
Detoxification of mercury (Hg) with selenium (Se) in the early postnatal period with regard to the expression of metallothionein protein (MT), essential element status, and lipid peroxidation level in tissues has not been studied. Seven-day-old Wistar pups were orally pretreated with Se [6 μmol Na2SeO3/kg body weight (b.w.)] for 3 days and then cotreated with Hg (6 μmol HgCl2/kg b.w.) for the following 4 days. This group (Se + Hg) was compared to the groups treated with Hg, Se, or vehicle (control). Compared to the Hg-group, Se + Hg-group exhibited lower renal MT expression, reduced accumulation of Hg, Cu and Zn, and reduced excretion of Se, Hg and Zn in urine. In the liver, MT was stimulated by Se treatment in both, Se and Se + Hg-group. Hepatic and brain levels of the endogenous essential elements Cu, Fe, Mg, and Zn remained unchanged in all of the studied groups. Brain Hg levels and oxidation of lipids measured as thiobarbituric acid reactive substances were diminished in Se + Hg-group of pups compared to the Hg-group. This study suggests that Se pretreatment can help reduce Hg in the tissues of suckling rats, simultaneously preventing impairment of essential element levels in the kidneys and their excessive excretion via urine. Also, Se was shown to prevent oxidative damage of lipids in the brain, which is particularly susceptible to Hg during the early postnatal period.
Sigma-Aldrich
Hydrogen chloride solution, 3 M in cyclopentyl methyl ether (CPME)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium selenite, 99%
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
D-Mannitol, BioUltra, ≥99.0% (sum of enantiomers, HPLC)
Sigma-Aldrich
Sodium selenite, γ-irradiated, lyophilized powder, BioXtra, suitable for cell culture
Sigma-Aldrich
D-Mannitol, ACS reagent
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
D-Mannitol, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
D-Mannitol, meets EP, FCC, USP testing specifications
Sigma-Aldrich
D-Mannitol, ≥98% (GC), suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
D-Mannitol, ≥98% (GC)
Sigma-Aldrich
2-Thiobarbituric acid, ≥98%
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium selenite, BioReagent, suitable for cell culture, ≥98%
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Hydrochloric acid, 36.5-38.0%, BioReagent, Molecular Biology
Sigma-Aldrich
Hydrochloric acid solution, 1.0 N, BioReagent, suitable for cell culture
Supelco
Hydrochloric acid solution, volumetric, 0.1 M HCl (0.1N), endotoxin free
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Hydrochloric acid solution, for amino acid analysis, ~6 M in H2O
Sigma-Aldrich
Hydrochloric acid solution, 32 wt. % in H2O, FCC
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)