Merck
  • Effects of the inhaled treatment of liriope radix on an asthmatic mouse model.
Effects of the inhaled treatment of liriope radix on an asthmatic mouse model.
The American journal of Chinese medicine (2015-05-15)
Ki-Suk Kim, Dong-Hyuk Cho, Hea Jung Yang, Eun-Kyeong Choi, Min Hee Shin, Kang-Hoon Kim, Kwang Seok Ahn, In Jin Ha, Yun-Cheol Na, Jae Young Um, Won Seok Chung, Hee-Jae Jung, Sung-Ki Jung, Hyeung-Jin Jang
ABSTRACT
As a treatment for allergic asthma, inhaled treatments such as bronchodilators that contain β2-agonists have an immediate effect, which attenuates airway obstructions and decreases airway hypersensitivity. However, bronchodilators only perform on a one off basis, but not consistently. Asthma is defined as a chronic inflammatory disease of the airways accompanying the overproduction of mucus, airway wall remodeling, bronchial hyperreactivity and airway obstruction. Liriope platyphylla radix extract (LPP), a traditional Korean medicine, has been thoroughly studied and found to be an effective anti-inflammatory medicine. Here, we demonstrate that an inhaled treatment of LPP can attenuate airway hyperresponsiveness (AHR) in an ovalbumin-induced asthmatic mouse model, compared to the saline-treated group (p < 0.01). Moreover, LPP decreases inflammatory cytokine levels, such as eotaxin (p < 0.05), IL-5 (p < 0.05), IL-13 (p < 0.001), RANTES (p < 0.01), and TNF-α (p < 0.05) in the bronchoalveolar lavage (BAL) fluid of asthmatic mice. A histopathological study was carried out to determine the effects of LPP inhalation on mice lung tissue. We performed UPLC/ESI-QTOF-MS, LC/MS, and GC/MS analyses to analyze the chemical constituents of LPP, finding that these are ophiopogonin D, spicatoside A, spicatoside B, benzyl alcohol, and 5-hydroxymethylfurfural. This study demonstrates the effect of an inhaled LPP treatment both on airway AHR and on the inflammatory response in an asthmatic mouse model. Hence, LPP holds significant promise as a nasal inhalant for the treatment of asthmatic airway disease.
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioXtra, ≥98.0%
Sigma-Aldrich
Aluminum potassium sulfate dodecahydrate, BioReagent, suitable for insect cell culture
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Hematoxylin
Sigma-Aldrich
Hematoxylin, certified by the BSC
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis