Merck
  • Islet transplantation from a nationally funded UK centre reaches socially deprived groups and improves metabolic outcomes.
Islet transplantation from a nationally funded UK centre reaches socially deprived groups and improves metabolic outcomes.
Diabetologia (2015-03-27)
Shareen Forbes, Neil W A McGowan, Kirsty Duncan, Debbie Anderson, Janet Barclay, Donna Mitchell, Kevin Docherty, David Turner, John D M Campbell, John J Casey
ABSTRACT
Type 1 diabetes complicated by hypoglycaemia is prevalent in socioeconomically deprived populations. Islet transplantation is of proven efficacy in type 1 diabetes complicated by hypoglycaemia, but it is not known if nationally funded programmes reach the socioeconomically deprived. Our aim was to determine: (1) socioeconomic indices in participants referred to our nationally funded programme; and (2) if metabolic outcomes in our transplant recipients were improved. Participants referred (n = 106) and receiving transplants (n = 18; 32 infusions) were examined with respect to socioeconomic status (deprivation category score) and their ability to work and drive. In participants followed for ≥12 months after transplantation, metabolic and anthropometric measurements (n = 14) were recorded pre- and post-transplant (assessed ~1, ~3, ~6 and ~12 months with mixed-meal tolerance tests and 6 day continuous glucose monitoring assessments). Donor data was also examined. There was a greater prevalence of socioeconomic deprivation in referred and transplant recipients than the general population (p < 0.05). Of the transplant recipients, 73% were socioeconomically deprived, 88% did not hold a driver's license and 94% had reduced ability to work (all p < 0.01 vs referred participants). Donors were predominantly obese and included circulatory death donors. At 12 months, 93% of participants who had received transplants had graft function, diminished frequency of hypoglycaemia (10 [4-11] vs 0 [0-2] hypoglycaemic episodes/week), improved awareness of hypoglycaemia (Gold score 7 [5-7] vs 1 [1-2]) and glycaemic control (HbA1c: 7.9% [7.2-8.5%]; 63 [55-69] mmol/mol vs 7.2% [6.8-7.5%]; 55 [51-58] mmol/mol), diminished glycaemic lability and decreased central adiposity (all p < 0.05). A nationally funded islet transplant programme reaches the socioeconomically deprived and outcomes are significantly improved in this group.
Sigma-Aldrich
DL-Alanine, ≥99%, FCC, FG
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
DL-Alanine, ≥99% (HPLC)
Sigma-Aldrich
Urea, BioXtra, pH 7.5-9.5 (20 °C, 5 M in H2O)
Supelco
Urea, 8 M (after reconstitution with 16 mL high purity water)
Sigma-Aldrich
Urea, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%, 99.0-101.0% (calc. on dry substance)
Sigma-Aldrich
Urea, ReagentPlus®, ≥99.5%, pellets
Sigma-Aldrich
Urea, suitable for electrophoresis
Sigma-Aldrich
Urea, ACS reagent, 99.0-100.5%
Sigma-Aldrich
Urea, powder, BioReagent, Molecular Biology, suitable for cell culture
Sigma-Aldrich
Urea, BioUltra, Molecular Biology, 99% (T)
Sigma-Aldrich
Urea, meets USP testing specifications
Sigma-Aldrich
Bilirubin, purum, ≥95.0% (UV)
Sigma-Aldrich
Urea, puriss. p.a., ACS reagent, reag. Ph. Eur., ≥99%
Sigma-Aldrich
Bilirubin, ≥98% (EmM/453 = 60), powder
Sigma-Aldrich
SyntheChol® NS0 Supplement, 500 ×, synthetic cholesterol, animal component-free, sterile-filtered, aqueous solution, suitable for cell culture
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C
Sigma-Aldrich
Urea-12C, 99.9 atom % 12C
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
Cholesterol, from sheep wool, ≥92.5% (GC), powder
Sigma-Aldrich
Cholesterol, Sigma Grade, ≥99%
Sigma-Aldrich
Cholesterol, powder, BioReagent, suitable for cell culture, ≥99%
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
Urea solution, 40 % (w/v) in H2O
Sigma-Aldrich
Urea solution, BioUltra, ~8 M in H2O
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra