Merck
  • Gradual Rewarming with Gradual Increase in Pressure during Machine Perfusion after Cold Static Preservation Reduces Kidney Ischemia Reperfusion Injury.
Gradual Rewarming with Gradual Increase in Pressure during Machine Perfusion after Cold Static Preservation Reduces Kidney Ischemia Reperfusion Injury.
PloS one (2015-12-03)
Paria Mahboub, Petra Ottens, Marc Seelen, Nils 't Hart, Nails t Hart, Harry Van Goor, Rutger Ploeg, Paulo N Martins, Paulo Martins, Henri Leuvenink
ABSTRACT
In this study we evaluated whether gradual rewarming after the period of cold ischemia would improve organ quality in an Isolated Perfused Kidney Model. Left rat kidneys were statically cold stored in University of Wisconsin solution for 24 hours at 4°C. After cold storage kidneys were rewarmed in one of three ways: perfusion at body temperature (38°C), or rewarmed gradually from 10°C to 38°C with stabilization at 10°C for 30 min and rewarmed gradually from 10°C to 38°C with stabilization at 25°C for 30 min. In the gradual rewarming groups the pressure was increased stepwise to 40 mmHg at 10°C and 70 mmHg at 25°C to counteract for vasodilatation leading to low perfusate flows. Renal function parameters and injury biomarkers were measured in perfusate and urine samples. Increases in injury biomarkers such as aspartate transaminase and lactate dehydrogenase in the perfusate were lower in the gradual rewarming groups versus the control group. Sodium re-absorption was improved in the gradual rewarming groups and reached significance in the 25°C group after ninety minutes of perfusion. HSP-70, ICAM-1, VCAM-1 mRNA expressions were decreased in the 10°C and 25°C groups. Based on the data kidneys that underwent gradual rewarming suffered less renal parenchymal, tubular injury and showed better endothelial preservation. Renal function improved in the gradual rewarming groups versus the control group.
Sigma-Aldrich
HEPES buffer solution, 1 M in H2O
Sigma-Aldrich
Creatinine, anhydrous, ≥98%
Sigma-Aldrich
4-Aminosalicylic acid, 99%
Sigma-Aldrich
Phenylacetic acid, ≥99%, FCC, FG
Sigma-Aldrich
Phenylacetic acid, 99%
Sigma-Aldrich
Bicinchoninic acid disodium salt hydrate, ≥98% (HPLC)
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ACS reagent, ≥99.0%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, ReagentPlus®, 99%
Sigma-Aldrich
1-Methyl-2-pyrrolidinone, biotech. grade, ≥99.7%
Sigma-Aldrich
HEPES, BioXtra, suitable for mouse embryo cell culture, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioXtra, pH 5.0-6.5 (1 M in H2O), ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioPerformance Certified, ≥99.5% (titration)
Sigma-Aldrich
HEPES, BioUltra, Molecular Biology, ≥99.5% (T)
Sigma-Aldrich
HEPES, ≥99.5% (titration)
Sigma-Aldrich
HEPES, anhydrous, free-flowing, Redi-Dri, ≥99.5%