Merck
  • New indole glucosides as biosynthetic intermediates of camptothecin from the fruits of Camptotheca acuminata.
New indole glucosides as biosynthetic intermediates of camptothecin from the fruits of Camptotheca acuminata.
Fitoterapia (2015-03-17)
Peng Wang, Jun Luo, Xiao-Bing Wang, Bo-Yi Fan, Ling-Yi Kong
ABSTRACT
Six new indole glucosides (1-6) and fifteen known alkaloids (7-21) were isolated from the fruit of Camptotheca acuminata. The planar structures of 1-6 were determined on the basis of spectroscopic data analysis and their absolute configurations were established by CD. The isolated indole glucosides showed a clear biosynthetic pathway of camptothecin (7), which started from tryptamine and secologanin and was proposed by synthetic chemists previously. Particularly, compound 1 supplemented the process of the transformation from pumiloside (20) or 3-epi-pumiloside (21) to camptothecin (7). In addition, camptothecin 10-O-β-D-glucopyranoside (13) and norcamptothecin (17), synthesized in the early structural modification of 7, were first isolated from the natural resources. The new compounds 1-6 were screened for their in vitro cytotoxicity but they did not show any exciting result.
Sigma-Aldrich
Dextrose, 97.5-102.0% anhydrous basis, meets EP, BP, JP, USP testing specifications
Sigma-Aldrich
D-(+)-Glucose, BioUltra, anhydrous, ≥99.5% (sum of enantiomers, HPLC)
Sigma-Aldrich
D-(+)-Glucose, ACS reagent
Sigma-Aldrich
D-(+)-Glucose, Hybri-Max, powder, BioReagent, suitable for hybridoma
Sigma-Aldrich
D-(+)-Glucose, suitable for mouse embryo cell culture, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, powder, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.5%
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC)
Sigma-Aldrich
D-(+)-Glucose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
D-Glucose-12C6, 16O6, 99.9 atom % 16O, 99.9 atom % 12C