Merck
  • Phytochemical Characterization of Chinese Bayberry (Myrica rubra Sieb. et Zucc.) of 17 Cultivars and Their Antioxidant Properties.
Phytochemical Characterization of Chinese Bayberry (Myrica rubra Sieb. et Zucc.) of 17 Cultivars and Their Antioxidant Properties.
International journal of molecular sciences (2015-06-05)
Xianan Zhang, Huizhong Huang, Qiaoli Zhang, Fangjuan Fan, Changjie Xu, Chongde Sun, Xian Li, Kunsong Chen
ABSTRACT
In order to fully understand the variations of fruit quality-related phytochemical composition in Chinese bayberry (Myrica rubra Sieb. et Zucc.), mature fruit of 17 cultivars from Zhejiang and Jiangsu provinces was used for the investigation of fruit quality attributes, including fruit color, soluble sugars, organic acids, total phenolics, flavonoids, antioxidant capacity, etc. Sucrose was the main soluble sugar, while citric acid was the main organic acid in bayberry fruit. The content of total phenolics and total flavonoids were positively correlated with 2,2-diphenyl-1-picrylhydrazyl (DPPH), ferric reducing antioxidant power (FRAP) antioxidant activity and 2,2'-azino-bis(3-ethylbenzothiazoline- 6-sulphonic acid) (ABTS) radical scavenging activity. Five anthocyanidins, i.e., delphinidin-hexoside (Dp-Hex), cyanidin-3-O-galactoside (C-3-Gal), cyanidin-3-O-glucoside (C-3-Glu), pelargonidin-3-O-glucoside (Pg-3-Glu) and peonidin-3-O-glucoside (Pn-3-Glu), and seven flavonols compounds, i.e., myricetin-3-O-rhamnoside (M-3-Rha), myricetin deoxyhexoside-gallate (M-DH-G), quercetin-3-O-galactoside (Q-3-Gal), quercetin-3- O-glucoside (Q-3-Glu), quercetin-3-O-rhamnoside (Q-3-Rha), kaempferol-3-O-galactoside (K-3-Gal) and kaempferol-3-O-glucoside (K-3-Glu), were identified and characterized among the cultivars. The significant differences in phytochemical compositions among cultivars reflect the diversity in bayberry germplasm, and cultivars of good flavor and/or rich in various health-promoting phytochemicals are good candidates for future genetic breeding of bayberry fruit of high quality. In conclusion, our results may provide important information for further breeding or industrial utilization of different bayberry resources.
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
2-Hydroxybutyric acid sodium salt, 97%
Sigma-Aldrich
DL-Malic acid, meets analytical specification of FCC, E296, 99-100.5% (alkalimetric)
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
2,2-Diphenyl-1-picrylhydrazyl
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, puriss., ≥99.5% (GC), meets analytical specification of Ph. Eur., BP, USP, crystalline (detached)
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
Sigma-Aldrich
DL-Malic acid, ≥98% (capillary GC)
Sigma-Aldrich
DL-Malic acid, 99%
Sigma-Aldrich
DL-Malic acid, ReagentPlus®, ≥99%