Merck
  • Abnormal Expression of miR-21 and miR-95 in Cancer Stem-Like Cells is Associated with Radioresistance of Lung Cancer.
Abnormal Expression of miR-21 and miR-95 in Cancer Stem-Like Cells is Associated with Radioresistance of Lung Cancer.
Cancer investigation (2015-04-02)
Jinghui Zhang, Chailan Zhang, Lin Hu, Yin He, Zeya Shi, Siyuan Tang, Yuxiang Chen
ABSTRACT
This study demonstrated that miR-21 and miR-95 expression were significantly higher in the ALDH1(+)CD133(+)subpopulation than in the ALDH1(-)CD133(-) subpopulation of lung cancer cells. Combined delivery of anti-miR-21 and anti-miR-95 by calcium phosphate nanoparticles significantly inhibited tumor growth in a xenograft tumor model and sensitized radiotherapy. The anti-miRNAs significantly reduced miR-21 and miR-95 levels, increased PTEN, SNX1, and SGPP1 protein expression, but reduced Akt Ser(473) and Thr(308) phosphorylation. ALDH1(+)CD133(+) subpopulation of NSCLC tumor cells confers radioresistance due to high expression of miR-21 and miR-95. Targeting inhibition of miR-21 and miR-95 can inhibit tumor growth through elevating PTEN, SNX1, and SGPP1 expression and inhibiting Akt phosphorylation.
Sigma-Aldrich
Iodonitrotetrazolium chloride, Used in colorimetric assays.
Sigma-Aldrich
Iodonitrotetrazolium chloride, BioReagent, ≥97.0% (calc. on dry substance, NT)
Sigma-Aldrich
Iodonitrotetrazolium chloride, 95%