Merck
  • Type 1 Insulin-Like Growth Factor Receptor/Insulin Receptor Substrate 1 Signaling Confers Pathogenic Activity on Breast Tumor Cells Lacking REST.
Type 1 Insulin-Like Growth Factor Receptor/Insulin Receptor Substrate 1 Signaling Confers Pathogenic Activity on Breast Tumor Cells Lacking REST.
Molecular and cellular biology (2015-06-24)
Kassondra Meyer, Brittany Albaugh, Barry Schoenike, Avtar Roopra
ABSTRACT
Loss of repressor element 1 silencing transcription factor (REST) occurs in 20% of breast cancers and correlates with a poor patient prognosis. However, the molecular basis for enhanced malignancy in tumors lacking REST (RESTless) is only partially understood. We used multiplatform array data from the Cancer Genome Atlas to identify consistent changes in key signaling pathways. Of the proteins screened in the reverse-phase protein array, we found that insulin receptor substrate 1 (IRS1) is the most highly upregulated protein in RESTless breast tumors. Analysis of breast tumor cell lines showed that REST directly represses IRS1, and cells lacking REST have increased levels of IRS1 mRNA and protein. We find that the upregulation of IRS1 function is both necessary and sufficient for enhanced signaling and growth in breast cancer cells lacking REST. IRS1 overexpression is sufficient to phenocopy the enhanced activation of the signaling hubs AKT and mitogen-activated protein kinase (MAPK) of MCF7 cells lacking REST. Loss of REST renders MCF7 and MDA-MB-231 breast tumor cells dependent on IRS1 activity for colony formation in soft agar. Inhibition of the type 1 insulin-like growth factor receptor (IGF1R) reduces the enhanced signaling, growth, and migration in breast tumor cells that occur upon REST loss. We show that loss of REST induces a pathogenic program that works through the IGF1R/IRS1 pathway.
Sigma-Aldrich
L-Glutamine
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Phenol, unstabilized, ReagentPlus®, ≥99%
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, 99.5-100.5% (GC)
Sigma-Aldrich
Phenol, puriss., meets analytical specification of Ph. Eur., BP, USP, ≥99.5% (GC), crystalline (detached)
Sigma-Aldrich
Sodium dodecyl sulfate solution, BioUltra, 20% in H2O
Sigma-Aldrich
Phenol, puriss. p.a., ACS reagent, reag. Ph. Eur., 99.0-100.5%
Sigma-Aldrich
Phenol, unstabilized, purified by redistillation, ≥99%
SAFC
L-Glutamine
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Phenol, BioUltra, Molecular Biology, TE-saturated, ~73% (T)
Sigma-Aldrich
Phenol, ≥99%
Sigma-Aldrich
Phenol, ACS reagent, ≥99.0%
Sigma-Aldrich
Sodium deoxycholate, BioXtra, ≥98.0% (dry matter, NT)
Sigma-Aldrich
Phenol, BioXtra, ≥99.5% (GC)
Sigma-Aldrich
Sodium deoxycholate, ≥97% (titration)
Sigma-Aldrich
Phenol, Molecular Biology
Sigma-Aldrich
Phenol solution, Saturated with 0.01 M citrate buffer, pH 4.3 ± 0.2, BioReagent, Molecular Biology
Sigma-Aldrich
Phenol solution, Equilibrated with 10 mM Tris HCl, pH 8.0, 1 mM EDTA, BioReagent, Molecular Biology
Sigma-Aldrich
Liquified Phenol, ≥89.0%
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
L-Glutamine, ReagentPlus®, ≥99% (HPLC)
Sigma-Aldrich
L-Glutamine, γ-irradiated, BioXtra, suitable for cell culture
Sigma-Aldrich
L-Glutamine, meets USP testing specifications, suitable for cell culture, 99.0-101.0%, from non-animal source
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Phenol, ≥96.0% (calc. on dry substance, T)
Sigma-Aldrich
MISSION® esiRNA, targeting mouse Irs1