Merck
  • Environmental friendly method for urban wastewater monitoring of micropollutants defined in the Directive 2013/39/EU and Decision 2015/495/EU.
Environmental friendly method for urban wastewater monitoring of micropollutants defined in the Directive 2013/39/EU and Decision 2015/495/EU.
Journal of chromatography. A (2015-10-04)
Ana R Ribeiro, Marta Pedrosa, Nuno F F Moreira, Manuel F R Pereira, Adrián M T Silva
ABSTRACT
The fate and removal of organic micropollutants in the environment is a demanding issue evidenced by the recent European policy. This work presents an analytical method for the trace quantification of 37 micropollutants including Priority Substances (Directive 2013/39/EU), substances of the recent watch list (Decision 2015/495/EU) and contaminants of emerging concern: pesticides, multi-class pharmaceuticals and a metabolite, estrogens and one industrial compound. The analytical method was based on solid phase extraction (SPE) followed by ultra-high performance liquid chromatography with tandem mass spectrometry (UHPLC-MS/MS), using eco-friendly solvents (ethanol and ultrapure water) both in the SPE and UHPLC, according to green analytical chemistry principles. Briefly, Oasis(®) HLB cartridges were used to preconcentrate 100mL of water samples and the reconstituted extracts were analyzed with a Kinetex™ column under reversed mode, the linear ranges extended to 200ngL(-1) (R(2)>0.99) for all the analytes. The method detection limits were between 0.04 and 2.26ngL(-1) and the method quantification limits were between 0.13 and 6.85ngL(-1). The identity of the compounds was confirmed using two MS/MS transitions and its ion ratios, according to Decision 2002/657/EC. The validated method was applied to wastewater treatment plant samples, assessing the concentration of micropollutants after secondary biological and tertiary UV treatments. Lab-scale photolysis and ozonation experiments were also performed with the secondary effluents, with ozonation showing the best performance for the removal of most of the determined micropollutants.
Sigma-Aldrich
Clarithromycin, ≥95% (HPLC)
Sigma-Aldrich
Clarithromycin, 96.0-102.0% (HPLC)
Sigma-Aldrich
Pentachlorophenol, 97%
Sigma-Aldrich
Ammonium acetate, 99.999% trace metals basis
Sigma-Aldrich
Sulfuric acid, 99.999%
Sigma-Aldrich
Carbamazepine, meets USP testing specifications
Sigma-Aldrich
Ammonium acetate, BioXtra, ≥98%
Sigma-Aldrich
Ammonium acetate, Molecular Biology, ≥98%
Sigma-Aldrich
Ammonium acetate, reagent grade, ≥98%
Sigma-Aldrich
Diclofenac sodium salt
Sigma-Aldrich
Ammonium acetate solution, Molecular Biology, 7.5 M
Sigma-Aldrich
(±)-Metoprolol (+)-tartrate salt, ≥98% (titration), powder
Sigma-Aldrich
Ibuprofen, ≥98% (GC)
Sigma-Aldrich
Diuron, ≥98%
Sigma-Aldrich
Bezafibrate, ≥98%, solid
Sigma-Aldrich
Trimethoprim, ≥98.5%
Sigma-Aldrich
Ibuprofen, meets USP testing specifications
Sigma-Aldrich
Trimethoprim, ≥99.0% (HPLC)
Sigma-Aldrich
Carbamazepine, powder
Sigma-Aldrich
Venlafaxine hydrochloride, ≥98% (HPLC), powder
Sigma-Aldrich
Methanol, Laboratory Reagent, ≥99.6%
Supelco
Ethanol solution, certified reference material, 2000 μg/mL in methanol
Supelco
Pentachlorophenol, analytical standard
Sigma-Aldrich
Sulfuric acid, puriss., meets analytical specification of Ph. Eur., BP, 95-97%
Supelco
Ibuprofen
Sigma-Aldrich
Methanol, ACS reagent, ≥99.8%
Sigma-Aldrich
Methanol, Absolute - Acetone free
Sigma-Aldrich
Methanol, suitable for NMR (reference standard)
Sigma-Aldrich
Sulfuric acid, ACS reagent, 95.0-98.0%
Sigma-Aldrich
Ammonium acetate, ACS reagent, ≥97%