Merck
  • The effect of skatole and androstenone on consumer response towards streaky bacon and pork belly roll.
The effect of skatole and androstenone on consumer response towards streaky bacon and pork belly roll.
Meat science (2015-07-18)
Margit D Aaslyng, Eva Honnens De Lichtenberg Broge, Per B Brockhoff, Rune Haubo Christensen
ABSTRACT
Consumer liking was assessed for streaky bacon and pork belly roll from entire male pigs with an androstenone (AND) content of up to 9.4 ppm and a skatole (SKA) content of up to 0.92 ppm in the back fat and castrates. No clear effect of either AND or SKA was seen in consumer liking, although an insignificant tendency was seen for SKA. A sensory profile analysis showed that AND increased the boar taint of bacon, while both AND and SKA increased the boar taint of the pork belly roll. Consumer sensitivity towards AND and SKA did not affect liking of the meat products. The lack of effect of AND and SKA on consumer liking could be due to a masking effect of the spices and smoke. Three consecutive weeks' exposure to bacon did not change the liking score, irrespective of the AND and SKA content. This indicates that the consumers did not become more sensitive towards boar taint.
Sigma-Aldrich
Dansylhydrazine, BioReagent, suitable for fluorescence, ≥90% (HPLC)
Sigma-Aldrich
Dansylhydrazine, 98%
Sigma-Aldrich
Potassium hydroxide, technical, ≥85%, powder
Sigma-Aldrich
Potassium hydroxide, reagent grade, 90%, flakes
Sigma-Aldrich
Potassium hydroxide, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, free-flowing, Redi-Dri, ACS reagent, ≥85%, pellets
Sigma-Aldrich
Potassium hydroxide, BioXtra, ≥85% KOH basis
Sigma-Aldrich
Potassium hydroxide, ≥85% KOH basis, pellets, white
Supelco
Potassium hydroxide solution, volumetric, 8.0 M KOH (8.0N)
Sigma-Aldrich
Potassium hydroxide, semiconductor grade, pellets, 99.99% trace metals basis (Purity excludes sodium content.)
Sigma-Aldrich
Potassium hydroxide solution, 45 wt. % in H2O
Sigma-Aldrich
1,2-Propanediol, ReagentPlus®, 99%
Sigma-Aldrich
1,2-Propanediol, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, meets analytical specification of Ph. Eur., BP, USP, ≥99.5%
Sigma-Aldrich
Acetonitrile, ReagentPlus®, 99%
Sigma-Aldrich
Potassium hydroxide, ≥99.95% trace metals basis
Sigma-Aldrich
Acetonitrile, ACS reagent, ≥99.5%
Sigma-Aldrich
1,2-Propanediol, puriss. p.a., ACS reagent, ≥99.5% (GC)
Sigma-Aldrich
Acetonitrile, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
Acetonitrile, anhydrous, 99.8%
Sigma-Aldrich
Acetonitrile, biotech. grade, ≥99.93%
Sigma-Aldrich
Acetonitrile, suitable for DNA synthesis, ≥99.9% (GC)
Sigma-Aldrich
Acetic acid-12C2, 99.9 atom % 12C
Sigma-Aldrich
Acetic acid, suitable for luminescence, BioUltra, ≥99.5% (GC)
Sigma-Aldrich
Acetic acid, ≥99.5%, FCC, FG
Sigma-Aldrich
Acetic acid, natural, ≥99.5%, FG
Sigma-Aldrich
5α-Androst-16-en-3-one
Sigma-Aldrich
Acetic acid, glacial, ACS reagent, ≥99.7%
Sigma-Aldrich
Acetic acid, glacial, ≥99.99% trace metals basis
Sigma-Aldrich
Acetic acid, glacial, ReagentPlus®, ≥99%