Merck
  • Quality improvement of transgenic cloned bovine embryos using an aggregation method: Effects on cell number, cell ratio, embryo perimeter, mitochondrial distribution, and gene expression profile.
Quality improvement of transgenic cloned bovine embryos using an aggregation method: Effects on cell number, cell ratio, embryo perimeter, mitochondrial distribution, and gene expression profile.
Theriogenology (2015-05-23)
J I Bang, J I Jin, N Ghanem, B H Choi, M Fakruzzaman, A N Ha, K L Lee, S J Uhm, D H Ko, B C Koo, J G Lee, I K Kong
ABSTRACT
The production of cloned embryos using conventional methods has extremely low success rates owing to low embryo quality. To improve the quality of cloned bovine embryos expressing enhanced green fluorescent protein (EGFP), we applied an aggregation culture method. The EGFP gene was transfected into bovine fetal fibroblasts using a retroviral vector system. Somatic cell nuclear transfer was performed using these cells, and the resulting embryos were cultured in aggregates or individually. Gene expression was analyzed by a microarray, and differentially expressed genes were validated by quantitative real-time polymerase chain reaction. The total number of cells per blastocyst and the ratio of inner cell mass cells to trophectoderm cells were higher in aggregated transgenic cloned blastocysts (agBL; 368.7 ± 109.6 and 1:4.8, respectively) than in in vitro-fertilized blastocysts (ivfBL; 189.8 ± 65.8 and 1:2.6, respectively) and nonaggregated transgenic cloned blastocysts (sBL; 113.1 ± 36.3 and 1:1.5, respectively; P < 0.05 and P < 0.01, respectively). Moreover, the blastocyst perimeter was larger in the agBL group than in the ivfBL and sBL groups (1168.8 ± 200.23 vs. 887.33 ± 187.62 and 678 ± 226.1 μm; P < 0.05). In addition, mitochondrial fluorescence intensity was higher in the agBL group than in the ivfBL and sBL groups (P < 0.05). The number of apoptotic cells per blastocyst was lower in the ivfBL and agBL groups than in the sBL group (3.7 ± 2.2 and 3.4 ± 2.1 vs. 6.7 ± 6.8; P < 0.05). The genes identified in the microarray belonged to 18 categories. Expression of the Krüppel-like factor 4 gene, which is associated with cell proliferation, development, and transcription, was 7.2-fold higher in the agBL group than in the ivfBL group (P < 0.05) but did not differ between the sBL and ivfBL groups (P > 0.05). Expression of the heat shock 70-kDa protein 1A gene, which is associated with apoptosis, was 12-fold higher in the sBL group than in the ivfBL and agBL groups (P < 0.05). Expression of a stemness-related gene (octamer-binding transcription factor 4) and trophectoderm-specific genes (homeobox protein CDX2 and keratin 18) was higher in the agBL group than in the sBL group (P < 0.05). However, expression of the stemness gene homeobox protein NANOG did not differ among the groups (P > 0.05). Taken together, these data suggest that the aggregation method improves the quality of cloned embryos expressing EGFP and might be helpful in animal cloning.
Sigma-Aldrich
Chloroform, anhydrous, contains amylenes as stabilizer, ≥99%
Sigma-Aldrich
Chloroform, ≥99%, PCR Reagent, contains amylenes as stabilizer
Sigma-Aldrich
Isopropyl alcohol, ≥99.7%, FCC, FG
Sigma-Aldrich
2-Propanol, electronic grade, 99.999% trace metals basis
Sigma-Aldrich
2-Propanol, HPLC Plus, for HPLC, GC, and residue analysis, 99.9%, poly coated bottles
Sigma-Aldrich
2-Propanol, BioReagent, ≥99.5%, Molecular Biology
Sigma-Aldrich
2-Propanol, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Fluorescein, free acid