Merck
  • Cysteine-free non-canonical C-intein for versatile protein C-terminal labeling through trans-splicing.
Cysteine-free non-canonical C-intein for versatile protein C-terminal labeling through trans-splicing.
Applied microbiology and biotechnology (2015-08-01)
Xudong Dai, Qijing Xun, Xiang-Qin Liu, Qing Meng
ABSTRACT
Site-specific protein labeling are powerful means of protein research and engineering; however, new and improved labeling methods are greatly needed. Split inteins catalyze a protein trans-splicing reaction that can be used for enzymatic and nearly seamless protein labeling. Non-canonical S11 split intein has been used in an earlier method of protein C-terminal labeling; however, its relatively large (~150 aa) N-intein fused to the target protein often hindered protein expression, folding, and solubility. To solve this problem, here, we have designed and demonstrated a new method of protein C-terminal labeling, by first engineering a functional non-canonical S1 split intein that has an extremely small (12 aa) N-intein and a cysteine-free C-intein. An engineered Rma DnaB S1 split intein was modified to have a cysteine-free C-intein, while still retaining its robust trans-splicing function, which permitted the C-extein in a C-precursor to have a single cysteine for easy and specific linkage with desired labeling groups. The resulting new and generally useful method has two unique advantages: (1) The extremely small (12 aa) N-intein, which must be fused to the C terminus of the target protein, is less likely to hinder the protein expression, folding, and solubility; and (2) the single cysteine in the C-extein may be readily linked to a variety of labeling or modification groups using commercially available reagents.
Sigma-Aldrich
Maltose solution, BioReagent, ~20% in H2O, Molecular Biology
Sigma-Aldrich
Potassium phosphate monobasic, BioUltra, Molecular Biology, anhydrous, ≥99.5% (T)
Sigma-Aldrich
Potassium phosphate monobasic, ReagentPlus®
Sigma-Aldrich
Potassium phosphate monobasic, Molecular Biology, ≥98.0%
Sigma-Aldrich
Potassium phosphate monobasic, 99.99% trace metals basis
Sigma-Aldrich
Potassium phosphate monobasic, powder, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99.0%
Sigma-Aldrich
Sodium chloride solution, 5 M in H2O, BioReagent, Molecular Biology
SAFC
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride, AnhydroBeads, −10 mesh, 99.999% trace metals basis
Sigma-Aldrich
DL-Dithiothreitol solution, BioUltra, Molecular Biology, ~1 M in H2O
Supelco
DL-Dithiothreitol solution, 1 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride, BioPerformance Certified, ≥99% (titration), suitable for insect cell culture, suitable for plant cell culture
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%