Merck
  • The Role of Janus Kinase 3 in the Regulation of Na⁺/K⁺ ATPase under Energy Depletion.
The Role of Janus Kinase 3 in the Regulation of Na⁺/K⁺ ATPase under Energy Depletion.
Cellular physiology and biochemistry : international journal of experimental cellular physiology, biochemistry, and pharmacology (2015-05-30)
Zohreh Hosseinzadeh, Sabina Honisch, Evi Schmid, Kashif Jilani, Kalina Szteyn, Shefalee Bhavsar, Yogesh Singh, Monica Palmada, Anja T Umbach, Ekaterina Shumilina, Florian Lang
ABSTRACT
Janus kinase-3 (JAK3) is activated during energy depletion. Energy-consuming pumps include the Na(+)/K(+)-ATPase. The present study explored whether JAK3 regulates Na(+)/K(+)-ATPase in dendritic cells (DCs). Ouabain (100 µM)-sensitive (Iouabain) and K(+)-induced (Ipump) outward currents were determined by utilizing whole cell patch-clamp, Na(+)/K(+)-ATPase α1-subunit mRNA levels by RT-PCR, Na(+)/K(+)-ATPase protein abundance by flow cytometry or immunofluorescence, and cellular ATP by luciferase-assay in DCs from bone marrow of JAK3-knockout (jak3(-/-)) or wild-type mice (jak3(+/+)). Ipump was further determined by voltage clamp in Xenopus oocytes expressing JAK3, active (A568V)JAK3 or inactive (K851A)JAK3. Na(+)/K(+)-ATPase α1-subunit mRNA and protein levels, as well as Ipump and Iouabain were significantly higher in jak3(-/-)DCs than in jak3(+/+)DCs. Energy depletion by 4h pre-treatment with 2,4-dinitro-phenol significantly decreased Ipump in jak3(+/+) DCs but not in jak3(-/-)DCs. Cellular ATP was significantly lower in jak3(-/-)DCs than in jak3(+/+)DCs and decreased in both genotypes by 2,4-dinitro-phenol, an effect significantly more pronounced in jak3(-/-)DCs than in jak3(+/+)DCs and strongly blunted by ouabain in both jak3(+/+) and jak3(-/-)DCs. Ipump and Iouabain in oocytes were decreased by expression of JAK3 and of (A568V)JAK3 but not of (K851A)JAK3. JAK3 inhibitor WHI-P154 (4-[(3'-bromo-4'-hydroxyphenyl)amino]-6,7-dimethoxyquinazoline, 22 μM) enhanced Ipump and Iouabain in JAK3 expressing oocytes. The difference between (A568V)JAK3 and (K851A)JAK3 expressing oocytes was virtually abrogated by actinomycin D (50 nM). JAK3 down-regulates Na(+)/K(+)-ATPase activity, an effect involving gene expression and profoundly curtailing ATP consumption.
Sigma-Aldrich
Sodium hydroxide-16O solution, 20 wt. % in H216O, 99.9 atom % 16O
Sigma-Aldrich
Sucrose, puriss., meets analytical specification of Ph. Eur., BP, NF
Sigma-Aldrich
Sucrose, BioUltra, Molecular Biology, ≥99.5% (HPLC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioXtra
Sigma-Aldrich
Sucrose, ≥99.5% (GC), BioReagent, suitable for cell culture, suitable for insect cell culture
Sigma-Aldrich
Sucrose, ACS reagent
Sigma-Aldrich
Sucrose, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, meets USP testing specifications
Sigma-Aldrich
Sucrose, ≥99% (GC), Grade I, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC), Grade II, suitable for plant cell culture
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sucrose, ≥99.5% (GC)
Sigma-Aldrich
Sodium chloride, 99.999% trace metals basis
Sigma-Aldrich
Sodium chloride, random crystals, 99.9% trace metals basis
Sigma-Aldrich
Sodium chloride-35Cl, 99 atom % 35Cl
Sigma-Aldrich
2,4-Dinitrophenol, moistened with water, ≥98.0%
Sigma-Aldrich
Sodium chloride solution, 0.85%
Sigma-Aldrich
Sodium hydroxide, BioUltra, suitable for luminescence, ≥98.0% (T), pellets
Sigma-Aldrich
Sodium chloride, BioUltra, Molecular Biology, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride solution, BioUltra, Molecular Biology, ~5 M in H2O
Sigma-Aldrich
Sodium chloride solution, 0.9% in water, BioXtra, suitable for cell culture
Sigma-Aldrich
Sodium chloride, BioReagent, suitable for cell culture, suitable for insect cell culture, suitable for plant cell culture, ≥99%
Sigma-Aldrich
Sodium chloride, BioXtra, ≥99.5% (AT)
Sigma-Aldrich
Sodium chloride, tablet
Sigma-Aldrich
Sodium chloride, meets analytical specification of Ph. Eur., BP, USP, 99.0-100.5%
Sigma-Aldrich
Sodium chloride, Molecular Biology, DNase, RNase, and protease, none detected, ≥99% (titration)
Sigma-Aldrich
Sodium chloride solution, 5 M
Sigma-Aldrich
Sodium hydroxide solution, 1.0 N, BioReagent, suitable for cell culture
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, Molecular Biology, ≥97.0%
Sigma-Aldrich
Ethylene glycol-bis(2-aminoethylether)-N,N,N′,N′-tetraacetic acid, BioXtra, ≥97 .0%