Merck
  • Liposomal thymoquinone effectively combats fluconazole-resistant Candida albicans in a murine model.
Liposomal thymoquinone effectively combats fluconazole-resistant Candida albicans in a murine model.
International journal of biological macromolecules (2015-02-25)
Masood Alam Khan, Ahmad N Aljarbou, Arif Khan, Hina Younus
ABSTRACT
The aim of the present study was to develop a novel liposomal formulation of thymoquinone (TQ) to treat fluconazole-susceptible and -resistant Candida albicans (C. albicans) infections. The liposomal preparation of TQ (Lip-TQ) was used against a fluconazole-susceptible or -resistant isolate of C. albicans. Various doses of fluconazole (0, 5, 10, 20 and 40 mg/kg) or free TQ or Lip-TQ (0, 1, 2 and 5mg/kg) were used to treat C. albicans infected mice. Mice were observed for 40 days post C. albicans infection, and their kidneys were assessed for the fungal load. Fluconazole showed anti-fungal activity against the drug-susceptible, but not against the -resistant isolate of C. albicans. Free TQ showed its activity against both fluconazole-susceptible or -resistant C. albicans, however, Lip-TQ was found to be the most effective and imparted ∼ 100% and ∼ 90% survival of mice infected with fluconazole-susceptible and -resistant isolates of C. albicans, respectively. Mice treated with Lip-TQ showed highly reduced severity of infection in their tissue homogenates. Therefore, Lip-TQ may effectively be used in the treatment of C. albicans infections, including those which are not responding to fluconazole.
Sigma-Aldrich
Dimethyl sulfoxide, Hybri-Max, sterile-filtered, BioReagent, suitable for hybridoma, ≥99.7%
Sigma-Aldrich
Dimethyl sulfoxide, meets EP testing specifications, meets USP testing specifications
Sigma-Aldrich
8-Octanoyloxypyrene-1,3,6-trisulfonic acid trisodium salt, suitable for fluorescence, ≥90% (HPCE)
Sigma-Aldrich
Dimethyl sulfoxide, BioUltra, Molecular Biology, ≥99.5% (GC)
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.5% (GC), suitable for plant cell culture
Sigma-Aldrich
Dimethyl sulfoxide, sterile-filtered, BioPerformance Certified, meets EP, USP testing specifications, suitable for hybridoma
Sigma-Aldrich
Dimethyl sulfoxide, Molecular Biology
Sigma-Aldrich
Dimethyl sulfoxide, PCR Reagent
Sigma-Aldrich
Dimethyl sulfoxide, ≥99.6%, ReagentPlus®
Sigma-Aldrich
Fluconazole, ≥98% (HPLC), powder
Sigma-Aldrich
MOPS, anhydrous, free-flowing, Redi-Dri, ≥99.5%
Sigma-Aldrich
MOPS, BioPerformance Certified, suitable for cell culture, ≥99.5% (titration)
Sigma-Aldrich
MOPS, BioXtra, ≥99.5% (titration)
Sigma-Aldrich
MOPS, ≥99.5% (titration)
Sigma-Aldrich
RPMI-1640 Medium, With L-glutamine and sodium bicarbonate, liquid, sterile-filtered, suitable for cell culture
Avanti
Cholesterol (plant), Avanti Research - A Croda Brand
Avanti
16:0 PC (DPPC), Avanti Research - A Croda Brand