Merck
  • Protein-Based Three-Dimensional Whispering-Gallery-Mode Micro-Lasers with Stimulus-Responsiveness.
Protein-Based Three-Dimensional Whispering-Gallery-Mode Micro-Lasers with Stimulus-Responsiveness.
Scientific reports (2015-08-05)
Yun-Lu Sun, Zhi-Shan Hou, Si-Ming Sun, Bo-Yuan Zheng, Jin-Feng Ku, Wen-Fei Dong, Qi-Dai Chen, Hong-Bo Sun
ABSTRACT
For the first time, proteins, a promising biocompatible and functionality-designable biomacromolecule material, acted as the host material to construct three-dimensional (3D) whispering-gallery-mode (WGM) microlasers by multiphoton femtosecond laser direct writing (FsLDW). Protein/Rhodamine B (RhB) composite biopolymer was used as optical gain medium innovatively. By adopting high-viscosity aqueous protein ink and optimized scanning mode, protein-based WGM microlasers were customized with exquisite true 3D geometry and smooth morphology. Comparable to previously reported artificial polymers, protein-based WGM microlasers here were endowed with valuable performances including steady operation in air and even in aqueous environments, and a higher quality value (Q) of several thousands (without annealing). Due to the "smart" feature of protein hydrogel, lasing spectrum was responsively adjusted by step of ~0.4 nm blueshift per 0.83-mmol/L Na2SO4 concentration change (0 ~ 5-mmol/L in total leading to ~2.59-nm blueshift). Importantly, other performances including Q, FWHM, FSR, peak intensities, exhibited good stability during adjustments. So, these protein-based 3D WGM microlasers might have potential in applications like optical biosensing and tunable "smart" biolasers, useful in novel photonic biosystems and bioengineering.
Sigma-Aldrich
Bovine Serum Albumin, lyophilized powder, essentially globulin free, ≥99% (agarose gel electrophoresis)